Z327271
BRAND® graduated cylinder, PMP, embossed scale
volume 25 mL, accuracy: 0.5 mL
Select a Size
$94.10
Estimated to ship onApril 17, 2025
Select a Size
About This Item
$94.10
Estimated to ship onApril 17, 2025
Recommended Products
Agency
ISO 6706 (meets DIN 12 680-1 tolerances for Class B cylinders)
feature
subdivision 0.5 mL
manufacturer/tradename
BRAND
volume measuring range
25 mL, accuracy: 0.5 mL
H
170 mm
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service