Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
graduated cylinder, PMP, embossed scale
Products
Product Category
cylinders (8)
Manufacturer
Agency
Height (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z327387
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 250 mL, accuracy: 2 mL
Expand
View Pricing
Z327484
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 2000 mL, accuracy: 20 mL
Expand
View Pricing
Z327352
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 100 mL, accuracy: 1 mL
Expand
View Pricing
Z327441
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 1000 mL, accuracy: 10 mL
Expand
View Pricing
Z327417
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 500 mL, accuracy: 5 mL
Expand
View Pricing
Z327247
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 10 mL, accuracy: 0.2 mL
Expand
View Pricing
Z327328
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 50 mL, accuracy: 1 mL
Expand
View Pricing
Z327271
BRAND
®
graduated cylinder, PMP, embossed scale,
volume 25 mL, accuracy: 0.5 mL
Expand
View Pricing