Skip to Content
Merck
All Photos(1)

Key Documents

X2879

Sigma-Aldrich

Xanthosine 5′-triphosphate triethylammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

150 UNITS
$389.00

$389.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
150 UNITS
$389.00

About This Item

Empirical Formula (Hill Notation):
C10H15N4O15P3 · xC6H15N
Molecular Weight:
524.17 (free acid basis)
UNSPSC Code:
41106305
NACRES:
NA.51

$389.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


storage temp.

−20°C

Quality Level

Related Categories

Physical form

Supplied as a 10 mM solution in water

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service