Skip to Content
Merck
All Photos(2)

Documents

X0750

Sigma-Aldrich

Xanthosine dihydrate

≥99%

Synonym(s):

9-β-D-Ribofuranosylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O6 · 2H2O
CAS Number:
Molecular Weight:
320.26
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99%

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless to faintly yellow

SMILES string

O.O.OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3C(=O)NC(=O)Nc23

InChI

1S/C10H12N4O6.2H2O/c15-1-3-5(16)6(17)9(20-3)14-2-11-4-7(14)12-10(19)13-8(4)18;;/h2-3,5-6,9,15-17H,1H2,(H2,12,13,18,19);2*1H2/t3-,5-,6-,9-;;/m1../s1

InChI key

ZCCPXSQIUORWCO-LGVAUZIVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthosine comprises xanthine ring. It is derived from a purine metabolite and is a caffeine biosynthesis intermediate.

Application

Xanthosine dihydrate has been used as a test compound in one dimensional liquid chromatography analysis (1D-LC ).

Biochem/physiol Actions

Xanthosine plays a key role in purine metabolism. High levels of xanthosine excretion is observed in breast cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive Natural Products II, 1, 977-1007 (2010)
Ewa Kulikowska et al.
Acta biochimica Polonica, 51(2), 493-531 (2004-06-26)
The 6-oxopurine xanthine (Xan, neutral form 2,6-diketopurine) differs from the corresponding 6-oxopurines guanine (Gua) and hypoxanthine (Hyp) in that, at physiological pH, it consists of a approximately 1:1 equilibrium mixture of the neutral and monoanionic forms, the latter due to
Robert W P Glowacki et al.
Cell host & microbe, 27(1), 79-92 (2020-01-07)
Efficient nutrient acquisition in the human gut is essential for microbial persistence. Although polysaccharides have been well-studied nutrients for the gut microbiome, other resources such as nucleic acids and nucleosides are less studied. We describe several ribose-utilization systems (RUSs) that
Ling Li et al.
Molecular cell, 80(3), 525-540 (2020-10-18)
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of
Lucas Willmann et al.
Scientific reports, 5, 13374-13374 (2015-08-22)
Cancer cells show characteristic effects on cellular turnover and DNA/RNA modifications leading to elevated levels of excreted modified nucleosides. We investigated the molecular signature of different subtypes of breast cancer cell lines and the breast epithelial cell line MCF-10A. Prepurification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service