Skip to Content
Merck
All Photos(1)

Documents

T46108

Sigma-Aldrich

1,2,4-Triazole

98%

Synonym(s):

3,4-Diazapyrrole, 4H-1,2,4-Triazole, s-Triazole (8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3N3
CAS Number:
Molecular Weight:
69.07
Beilstein:
104767
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

260 °C (lit.)

mp

119-121 °C (lit.)

SMILES string

c1nc[nH]n1

InChI

1S/C2H3N3/c1-3-2-5-4-1/h1-2H,(H,3,4,5)

InChI key

NSPMIYGKQJPBQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,4-triazole and its derivatives are important structural moieties of many pharmaceutical drugs. Triazoles can also act as ligands to form coordination complexes with transition metal ions. Due to their electron-deficient nature, they exhibit excellent electron-transport and hole-blocking properties, making them promising organic materials in material science applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mononuclear, oligonuclear and polynuclear metal coordination compounds with 1, 2, 4-triazole derivatives as ligands.
Haasnoot JG
Coordination Chemistry Reviews, 200, 131-185 (2000)
Applications of Metal-Free 1, 2, 4-Triazole Derivatives in Materials Science.
Diaz-Ortiz A
Current Organic Chemistry, 19(7), 568-584 (2015)
1, 2, 4-Triazoles: Synthetic approaches and pharmacological importance.
Al-Masoudi IA
Chemistry of Heterocyclic Compounds, 42(11), 1377-1403 (2006)
Tomasz Plech et al.
European journal of medicinal chemistry, 60, 128-134 (2013-01-05)
A series of novel 1,2,4-triazole-ciprofloxacin hybrids was designed, synthesised and evaluated in vitro against drug-susceptible and drug-resistant bacteria. A significant part of the compounds obtained showed antibacterial activity higher than the activity of ciprofloxacin, both towards Gram-positive and Gram-negative species.
Camila C Amorim et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(3), 183-190 (2013-01-30)
This study evaluated the adsorption capacity of ethylenthiourea (ETU) and 1H-1,2,4-triazole (1,2,4-T) for two commercial activated carbons: charcoal-powdered activated carbon (CPAC) and bovine bone-powdered activated carbon (BPAC). The tests were conducted at a bench scale, with ETU and 1,2,4-T diluted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service