Key Documents
V5382
Nalgene® specimen vial
capacity 15 mL
Synonym(s):
sample container, sample vial, specimen container
Select a Size
1 PKG
$245.00
$245.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 PKG
$245.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$245.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
material
high-density polyethylene cap
polypropylene copolymer bottle
round translucent bottle
Joint
joint threaded neck
sterility
sterile
feature
with cap
packaging
pack of 75 ea
manufacturer/tradename
Nalgene® 5005-0015
capacity
15 mL
General description
Vials of polypropylene, linerless screw closures of high-density polyethylene. For storage of solid specimens. Use in mechanical freezers only. Wide-mouth, shoulderless vial allows easy access to sample. Flat bottom permits self-standing on flat surface. Certified sterile, non-cytotoxic and non-pyrogenic.
Packaging
75 vials per package
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)