Skip to Content
Merck
All Photos(1)

Key Documents

V5257

Sigma-Aldrich

Nalgene® cryogenic vials

capacity 5 mL, sterile

Synonym(s):

cryostorage vial, cryovial cryotube

Sign Into View Organizational & Contract Pricing

Select a Size

250 EA
$466.00

$466.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
250 EA
$466.00

About This Item

UNSPSC Code:
41121704
NACRES:
NB.22

$466.00


Estimated to ship onMay 21, 2025


material

colorless tube
conical bottom
high-density polyethylene closure
polypropylene
polypropylene tube

sterility

sterile

feature

External thread

packaging

pack of 250 ea (25 x 10 ea)

manufacturer/tradename

Nalgene® 5000-0050

parameter

-150 °C min. temp.

technique(s)

cryopreservation: suitable

capacity

5 mL

tube diam.

12.7 mm

Looking for similar products? Visit Product Comparison Guide

General description

Vials designed for exceptional ease of use and sample integrity in ultra-low temperature storage. Closure seals without gasket or O-ring, minimizing risk of contamination. The deep skirt of the closure allows single-handed aseptic technique without exposing fingers to vial contents. Ribs around the bottom of the vial interlock with wells in the Cryovial Racks so the vial will not turn when opening or closing. Bottoms of vials are skirted; vial stands without holder.

Sterile vials and closures are pre-assembled and radiation-sterilized. A white marking area, fill line and graduations are printed on the side. Certified sterile, non-cytotoxic and non-pyrogenic.

Note: Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex tubing. Such use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up resulting in possible explosion or biohazard release.

Legal Information

CryoFlex is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service