Skip to Content
Merck
All Photos(1)

Key Documents

Z740433

HistoTainer II prefilled specimen containers

M961, volume 90 mL

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
$199.00

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
96 EA
$199.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polyethylene cap
polypropylene container

packaging

pack of 96 ea

manufacturer/tradename

Simport M961-90FMA

volume

90 mL

Looking for similar products? Visit Product Comparison Guide

General description

NON Tamper Evident Prefilled Specimen Containers, 50% filled with 10% Neutral Buffered Formalin

Especially designed for collection, transport and storage of histology specimens, Simport offers shatter resistant polypropylene containers, eliminating most problems of leakage and evaporation. Containers are manufactured from virgin, translucent polypropylene. The white lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable for easy, safe storage. Jars are translucent and specimens can be viewed without having to open the lid. Closures are manufactured from virgin polyethylene with a unique integrated leak-resistant seal.

Both containers and caps are manufactured without the use of plasticisers or mold release agents. All materials used in manufacturing are free from latex. Containers are
95 kPa compliant. Available in many sizes from 20 to 120 ml. Packaging is offered in trays of 24, cases of 96.

The HistoTainer II is half filled with a choice of fixatives. 10% Neutral Buffered Formalin penetrates quickly, but fixes slowly. The Formalin is enhanced by a buffering capacity optimizing histological results by light microscopy and immunohistochemistry. Bouin, Hollande and Zinc Formalin fixatives are also available.

Legal Information

HistoTainer is a trademark of Simport

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service