Skip to Content
Merck
All Photos(1)

Key Documents

U8879

Sigma-Aldrich

Hydrogen peroxide–Urea adduct

tablet

Synonym(s):

Carbamide Per hydrate, Carbamide peroxide, Percarbamide, Urea hydrogen peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(NH2)2 · H2O2
CAS Number:
Molecular Weight:
94.07
Beilstein:
3680414
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

vapor pressure

23.3 mmHg ( 30 °C)

form

tablet

reaction suitability

reagent type: oxidant

mp

90-93 °C (lit.)

solubility

water: 1 tablets/4 mL, clear, colorless

storage temp.

2-8°C

SMILES string

OO.NC(N)=O

InChI

1S/CH4N2O.H2O2/c2-1(3)4;1-2/h(H4,2,3,4);1-2H

InChI key

AQLJVWUFPCUVLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

Urea Hydrogen Peroxide Tablets provide a safe and convenient source of H2O2 for use in enzyme immunoassay and other procedures.

Quantity

One tablet will provide the equivalent of 1.75 mg of H2O2.

Reconstitution

When dissolved in 12.5 mL deionized water, one tablet yields a solution containing 0.014% H2O2.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Cavalli et al.
Brazilian oral research, 26(6), 536-542 (2012-11-28)
This study evaluated the influence of fluoride-containing carbamide peroxide (CP) bleaching agents and adhesive systems on bonded enamel interfaces that are part of the dynamic pH cycling and thermal cycling models. The buccal surfaces of 60 bovine incisors were restored
Lidia Yileng Tay et al.
American journal of dentistry, 25(4), 199-204 (2012-10-23)
This parallel, double-blind randomized clinical trial evaluated the 2-year bleaching efficacy and sensitivity produced by at-home (AH) and in-office (IO) bleaching therapies. 60 participants with tooth color darker than C2, without restorations in the anterior dentition and older than 18
Karen Pintado-Palomino et al.
Journal of dentistry, 43(9), 1099-1105 (2015-07-15)
To evaluate the efficacy of experimental proposals of desensitizing agents during tooth bleaching. 140 participants without tooth sensitivity (TS) received 16% carbamide peroxide (14 days-04 h each) (T1) or 35% hydrogen peroxide (single session-45 min) (T2). Participants used concomitantly (10
L E Tam et al.
Operative dentistry, 38(2), 142-150 (2012-09-01)
Although damage to the structural integrity of the tooth is not usually considered a significant problem associated with tooth bleaching, there have been some reported negative effects of bleaching on dental hard tissues in vitro. More studies are needed to
Hsin H Kuo et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(35), 13966-13971 (2012-08-15)
The heme enzyme indoleamine 2,3-dioxygenase (IDO) was found to catalyze the oxidation of indole by H(2)O(2), with generation of 2- and 3-oxoindole as the major products. This reaction occurred in the absence of O(2) and reducing agents and was not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service