Skip to Content
Merck
All Photos(1)

Key Documents

P9187

Sigma-Aldrich

OPD

SIGMAFAST, peroxidase substrate, chromogenic, tablet

Synonym(s):

OPD Peroxidase Substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

Product Name

SIGMAFAST OPD, tablet

form

tablet

Quality Level

solubility

water: 0.5 tablet/10 mL, clear, colorless

storage temp.

2-8°C

Application

SIGMAFAST OPD (o-Phenylenediamine dihydrochloride) tablets have been developed for use as a soluble substrate for the detection of peroxidase activity in enzyme immunoassays (EIA). OPD is the EIA substrate of choice as it exhibits high sensitivity. EIA applications utilizing OPD may be read in timed assays or stopped with dilute acid solutions for delayed readings.
SIGMAFAST OPD has been used in saliva- virus-like particles (VLP) binding assay. It has also been used to develop enzyme linked immunosorbent assay (ELISA) plates for the detection of rituximab in rat serum samples.

Features and Benefits

SIGMAFAST OPD Tablets require no additional buffers or steps to prepare an active substrate solution.

Caution

This substrate is hygroscopic and light sensitive

Reconstitution

Each tablet set dissolved in 20 mL deionized water yields a ready-to-use buffered solution containing OPD and urea hydrogen peroxide.
One OPD tablet and one urea hydrogen peroxide/buffer tablet, dissolved in 20 ml of water, provides 20 ml of ready-to-use substrate. When the tablet is dissolved in this manner, the final concentrations will be 0.4 mg/ml OPD, 0.4 mg/ml urea hydrogen peroxide, and 0.05 M phosphate-citrate, pH 5.0. Tablets are individually packaged in foil packets.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subcutaneous absorption of monoclonal antibodies: role of dose, site of injection, and injection volume on rituximab pharmacokinetics in rats
Kagan L, et al.
Pharmaceutical Research, 29(2), 490-499 (2012)
Elisa Lobato et al.
Ecology and evolution, 7(11), 3645-3656 (2017-06-16)
Oceanic islands share several environmental characteristics that have been shown to drive convergent evolutionary changes in island organisms. One change that is often assumed but has seldom been examined is the evolution of weaker immune systems in island species. The
Courtney Woolsey et al.
Nature immunology, 22(1), 86-98 (2020-11-26)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is responsible for an unprecedented global pandemic of COVID-19. Animal models are urgently needed to study the pathogenesis of COVID-19 and to screen vaccines and treatments. We show that African green monkeys (AGMs)
Jeffrey D Whitman et al.
Nature biotechnology, 38(10), 1174-1183 (2020-08-29)
Appropriate use and interpretation of serological tests for assessments of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) exposure, infection and potential immunity require accurate data on assay performance. We conducted a head-to-head evaluation of ten point-of-care-style lateral flow assays (LFAs)
Almahamoudou Mahamar et al.
Malaria journal, 16(1), 289-289 (2017-07-20)
Seasonal malaria chemoprevention (SMC) is a new strategy to reduce malaria burden in young children in Sahelian countries. It consists of the administration of full treatment courses of sulfadoxine-pyrimethamine plus amodiaquine to children at monthly intervals during the malaria season.

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Protocols

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service