Skip to Content
Merck
All Photos(1)

Key Documents

U4626

Sigma-Aldrich

Undecanoyl chloride

~99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9COCl
CAS Number:
Molecular Weight:
204.74
Beilstein:
1759226
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

refractive index

n20/D 1.443 (lit.)

bp

135-136 °C/20 mmHg (lit.)

density

0.93 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCC(Cl)=O

InChI

1S/C11H21ClO/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3

InChI key

JUKPJGZUFHCZQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service