Key Documents
Z129607
Sigma-Aldrich
Aldrich® micro distillation head
joint: ST/NS 24/40
Select a Size
1 EA
$615.00
$615.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$615.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$615.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
description
glass stopcock plug 2 mm with O-ring
Related Categories
General description
Use with recovery flask Z515574.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)