Skip to Content
Merck
All Photos(2)

Key Documents

S2GPT05RE

Millipore

Millipore® Steritop® Vacuum Bottle Top Filter

pore size 0.22 μm, polyethersulfone membrane, funnel capacity 500 mL, pack of 12 ea

Synonym(s):

SCGPT05RE, Steritop® QR Quick Release Bottle Top Filter, bottle top sterile filter unit, bottle top vacuum filter, bottle top vacuum filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24
pore size:
0.22 μm pore size
H:
148 mm
capacity:
500 mL
diam.:
12.5 cm
disc diam.:
73 mm
filter diam.:
73 mm
filtration area:
40 cm2
funnel capacity:
500 mL
material:
polyethersulfone membrane
polystyrene funnel(s)
polystyrene housing
manufacturer/tradename:
Steritop®

material

polyethersulfone membrane
polystyrene funnel(s)
polystyrene housing

Quality Level

sterility

sterile; irradiated

product line

Steritop®

feature

hydrophilic

packaging

pack of 12 ea

manufacturer/tradename

Steritop®

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

148 mm

capacity

500 mL

diam.

12.5 cm

disc diam.

73 mm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

500 mL

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel
45 mm outlet thread(s)

shipped in

ambient

General description

This item is a direct replacement for SCGPT05RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Steritop® 500 mL bottle-top with 0.22 μm pore size polyethersulfone (PES) membrane filter unit can be used on bottles with 45 mm openings, designed for vacuum applications.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Evaluation of sterile filtration devices for protein preservation in sterile filtration of cell culture media.

Related Content

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

This article shows that a PES membrane containing Stericup Quick Release filter provides an alternative for the pluripotent expansion of embryonic stem cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service