Skip to Content
Merck
All Photos(1)

Key Documents

TMO276005

Nunc® 96 DeepWell plate well cap

chemically resistant, pierceable well cap mats, blue, non-sterile, case of 50 ea

Synonym(s):

plate cover, plate cover mat

Sign Into View Organizational & Contract Pricing

Select a Size

50 EACH
$513.00

$513.00


Estimated to ship onMay 25, 2025



Select a Size

Change View
50 EACH
$513.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

$513.00


Estimated to ship onMay 25, 2025


sterility

non-sterile

packaging

case of 50 ea

manufacturer/tradename

Nunc 276005

color

blue

General description

Protect all 96 wells during short-term or long-term storage of chemicals or compounds with a single device using Nunc™ 96-Well Cap Mats, which prevent contamination and evaporation of well contents.

Features and Benefits

  • Chemically resistant
  • Pierceable
  • Can withstand low temperature to -80°C

Legal Information

DeepWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service