Skip to Content
Merck
All Photos(1)

Key Documents

SRP3031

Sigma-Aldrich

Endostatin human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

COL18A1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$453.00

$453.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
$453.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$453.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

20.2 kDa

packaging

pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... COL18A1(80781)

General description

Research area: Cell Signaling

Endostatin is an endogenous glycoprotein derived from the C-terminal portion of the α1 chain of type XVIII collagen. [1] It is encoded by the gene mapped to human chromosome 21q22.3. Recombinant human Endostatin protein consists of 184 amino acid residues. It is expressed in various organs with the highest levels in liver, lung, and kidney. [2][3]

Biochem/physiol Actions

Endostatin is a potent inhibitor of angiogenesis. It exerts its angiostatic effect through multiple pathways related to endothelial cell migration and proliferation. Endostatin inhibits and suppresses the actions of matrix metalloproteinases (MMPs), FAK (Focal adhesion kinase)/Ras/p38-MAPK (mitogen-activated protein kinase)/ERK (extracellular signal-regulated kinase) signaling cascade, HIF-1α(hypoxia-inducible factor 1 alpha)/VEGF-A (vascular endothelial growth factor)thereby inhibiting angiogenesis by preventing the migration and invasion of endothelial cells.[4] Endostatin also blocks the apoptosis of cells and the G1 to S phase transition in the cell cycle. Mutations in this gene lead to Knobloch syndrome (KS), a rare autosomal recessive disorder.[2][5]

Sequence

MHSHRDFQPV LHLVALNSPL SGGMRGIRGA DFQCFQQARA VGLAGTFRAF LSSRLQDLYS IVRRADRAAV PIVNLKDELL FPSWEALFSG SEGPLKPGAR IFSFDGKDVL RHPTWPQKSV WHGSDPNGRR LTESYCETWR TEAPSATGQA SSLLGGRLLG QSAASCHHAY IVLCIENSFM TASK

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Digtyar et al.
Biochemistry. Biokhimiia, 72(3), 235-246 (2007-04-24)
Endogenous inhibitors of angiogenesis are proved to be a major factor preventing the emergence of clinically manifested stages of human cancer. The protein endostatin, a 20-kD proteolytic fragment of type XVIII collagen, is one of the most active natural inhibitors
S P Oh et al.
Genomics, 19(3), 494-499 (1994-02-01)
Types XV and XVIII collagen belong to a unique and novel subclass of the collagen superfamily for which we have proposed the name the MULTIPLEXIN family. Members of this class contain polypeptides with multiple triple-helical domains separated and flanked by
Extracellular matrix modulates angiogenesis in physiological and pathological conditions.
Neve A
BioMed Research International, 756078-756078 (2014)
Maria Rita Passos-Bueno et al.
Anais da Academia Brasileira de Ciencias, 78(1), 123-131 (2006-03-15)
Collagen XVIII, a proteoglycan, is a component of basement membranes (BMs). There are three distinct isoforms that differ only by their N-terminal, but with a specific pattern of tissue and developmental expression. Cleavage of its C-terminal produces endostatin, an inhibitor
Ilkka Alahuhta et al.
Experimental cell research, 336(1), 130-140 (2015-06-27)
The turnover of extracellular matrix liberates various cryptic molecules with novel biological activities. Endostatin is an endogenous angiogenesis inhibitor that is derived from the non-collagenous domain of collagen XVIII. Although there are a large number of studies on its anti-tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service