Key Documents
BR701368
BRAND® cover mat for 96-well plates, deep-well
2 mL low profile, TPE, non-sterile
Synonym(s):
Accessories, Cover mats, Deep-well Plates, Plates, Sample Storage
Select a Size
50 EA
$472.00
$472.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
50 EA
$472.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
TPE
size:
96 well
$472.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
TPE
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701368
size
96 well
General description
These mats provide a reliable barrier to minimize evaporation and contamination during experiments. Cover mats reduce the maximum volume of wells. Adhesive sealing films can also be used.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)