Skip to Content
Merck
All Photos(1)

Key Documents

T3450

Sigma-Aldrich

Thiolutin

from Streptomyces luteosporeus, ≥95% (HPLC)

Synonym(s):

Farcinicin, N-(4,5-Dihydro-4-methyl-5-oxo-1,2-dithiolo[4,3-b]pyrrol-6-yl), Propiopyvothine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$495.00

$495.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$495.00

About This Item

Empirical Formula (Hill Notation):
C8H8N2O2S2
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352105
PubChem Substance ID:
NACRES:
NA.77

$495.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

biological source

Streptomyces luteosporeus

Quality Level

Assay

≥95% (HPLC)

form

solid

solubility

DMSO: 0.90 - 1.10 mg/ml, clear, yellow

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCC(=O)NC1=C2SSC=C2N(C)C1=O

InChI

1S/C9H10N2O2S2/c1-3-6(12)10-7-8-5(4-14-15-8)11(2)9(7)13/h4H,3H2,1-2H3,(H,10,12)

InChI key

UGZYFXMSMFMTSM-UHFFFAOYSA-N

Application

Thiolutin has been used as a polymerase II inhibitor:
  • to study its effects on yeast cells to calculate transcript half-life [1]
  • to study its effects on transcription during germination in budding yeast [2]
  • to study its effects on cell adhesion in zebrafish [3]

Biochem/physiol Actions

Sulfur-containing antibiotic, which is a potent inhibitor of bacterial and yeast RNA polymerases. It was found to inhibit in vitro RNA synthesis directed by all three yeast RNA polymerases (I, II, and III). Thiolutin is also an inhibitor of mannan and glucan formation in Saccharomyces cerevisiae and used for the analysis of mRNA stability. Studies have shown that thiolutin inhibits adhesion of human umbilical vein endothelial cells (HUVECs) to vitronectin and thus suppresses tumor cell-induced angiogenesis in vivo.

Preparation Note

Thiolutin dissolves in DMSO at 0.90 - 1.10 mg/ml to yield a clear, yellow solution.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kadowaki et al.
The Journal of cell biology, 126(3), 649-659 (1994-08-01)
To understand the mechanisms of mRNA transport in eukaryotes, we have isolated Saccharomyces cerevisiae temperature-sensitive (ts) mutants which accumulate poly(A)+ RNA in the nucleus at the restrictive temperature. A total of 21 recessive mutants were isolated and classified into 16
Qiaoning Guan et al.
PLoS genetics, 2(11), e203-e203 (2006-12-15)
Nonsense-mediated mRNA decay (NMD) is a eukaryotic mechanism of RNA surveillance that selectively eliminates aberrant transcripts coding for potentially deleterious proteins. NMD also functions in the normal repertoire of gene expression. In Saccharomyces cerevisiae, hundreds of endogenous RNA Polymerase II
Thiolutin inhibits endothelial cell adhesion by perturbing Hsp27 interactions with components of the actin and intermediate filament cytoskeleton
Jia Y, et al.
Cell Stress & Chaperones, 15(2), 165-181 (2010)
Size-dependent expression of the mitotic activator Cdc25 suggests a mechanism of size control in fission yeast
Keifenheim D, et al.
Current Biology, 27(10), 1491-1497 (2017)
Brett N Tomson et al.
Molecular and cellular biology, 26(16), 6239-6247 (2006-08-02)
The ribosomal DNA (rDNA) is a specialized genomic region not only owing to its function as the nucleolar organizing region (NOR) but also because it is repetitive in nature and, at least in budding yeast, silenced for polymerase II (Pol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service