Skip to Content
Merck
All Photos(3)

Key Documents

605573

Sigma-Aldrich

Palmitic acid-13C16

99 atom % 13C, 99% (CP)

Synonym(s):

Hexadecanoic acid-13C16

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$2,350.00

$2,350.00


Estimated to ship onMay 29, 2025


Request more information

Select a Size

Change View
100 MG
$2,350.00

About This Item

Linear Formula:
13CH3(13CH2)1413CO2H
CAS Number:
Molecular Weight:
272.31
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$2,350.00


Estimated to ship onMay 29, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

mp

61-64 °C

mass shift

M+16

storage temp.

room temp

SMILES string

[13CH3][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1

InChI key

IPCSVZSSVZVIGE-BZDICNBSSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

402.8 °F - closed cup

Flash Point(C)

206 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Chen et al.
Gastroenterology, 158(4), 985-999 (2019-11-25)
Functions of intestinal stem cells (ISCs) are regulated by diet and metabolic pathways. Hepatocyte nuclear factor 4 (HNF4) family are transcription factors that bind fatty acids. We investigated how HNF4 transcription factors regulate metabolism and their functions in ISCs in
Matteo Audano et al.
The Journal of cell biology, 220(3) (2021-02-11)
The commitment of mesenchymal stem cells to preadipocytes is stimulated by hormonal induction. Preadipocytes induced to differentiate repress protein synthesis, remodel their cytoskeleton, and increase mitochondrial function to support anabolic pathways. These changes enable differentiation into mature adipocytes. Our understanding
Riccardo M Betto et al.
Nature genetics, 53(2), 215-229 (2021-02-03)
Naive epiblast and embryonic stem cells (ESCs) give rise to all cells of adults. Such developmental plasticity is associated with genome hypomethylation. Here, we show that LIF-Stat3 signaling induces genomic hypomethylation via metabolic reconfiguration. Stat3-/- ESCs show decreased α-ketoglutarate production
M Zaeem Cader et al.
Cell, 180(2), 278-295 (2020-01-25)
Mutations in FAMIN cause arthritis and inflammatory bowel disease in early childhood, and a common genetic variant increases the risk for Crohn's disease and leprosy. We developed an unbiased liquid chromatography-mass spectrometry screen for enzymatic activity of this orphan protein.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service