Skip to Content
Merck
All Photos(3)

Key Documents

I7017

Sigma-Aldrich

Indole-3-pyruvic acid

≥97%

Synonym(s):

3-(3-Indolyl)-2-oxopropanoic acid, 3-Indolylpyruvic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$259.00
5 G
$1,230.00

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$259.00
5 G
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Beilstein:
172966
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

color

light yellow

mp

215 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC(=O)C(=O)Cc1c[nH]c2ccccc12

InChI

1S/C11H9NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5H2,(H,14,15)

InChI key

RSTKLPZEZYGQPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Indole-3-pyruvic acid can be used:
  • As a precursor for the synthesis of chromopyrrolic acid by a heme-containing enzyme.[1]
  • As a reactant in the Biginelli-like scaffold syntheses.[2]

Linkage

α-Keto analogue of tryptophan

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Bacciottini et al.
Pharmacological research communications, 19(11), 803-817 (1987-11-01)
The effects of acute or repeated administration of indole-pyruvic acid (IPA), a keto-analogue of tryptophan (TRP), were studied in various brain areas of rats by measuring the changes of 5-hydroxytryptamine (5-HT) and of norepinephrine (NE) content and metabolism. The analgesic
Cyclic ketones and substituted α-keto acids as alternative substrates for novel Biginelli-like scaffold syntheses
Abelman, Matthew M, et al.
Tetrahedron Letters, 44(24), 4559-4562 (2003)
Direct formation of chromopyrrolic acid from indole-3-pyruvic acid by StaD, a novel hemoprotein in indolocarbazole biosynthesis
Asamizu, S, et al.
Tetrahedron Letters, 47(4), 473-475 (2006)
Nathan D Tivendale et al.
Plant physiology, 159(3), 1055-1063 (2012-05-11)
Seeds of several agriculturally important legumes are rich sources of the only halogenated plant hormone, 4-chloroindole-3-acetic acid. However, the biosynthesis of this auxin is poorly understood. Here, we show that in pea (Pisum sativum) seeds, 4-chloroindole-3-acetic acid is synthesized via
Goutam Chowdhury et al.
Chemical research in toxicology, 22(12), 1905-1912 (2009-10-29)
Aerobic incubation of the tryptophan transamination/oxidation product indole-3-pyruvic acid (I3P) at pH 7.4 and 37 degrees C yielded products with activity as Ah receptor (AHR) agonists. The extracts were fractionated using HPLC and screened for AHR agonist activity. Two compounds

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service