Skip to Content
Merck
All Photos(1)

Key Documents

SRP8050

Sigma-Aldrich

Profilin from Toxoplasma gondii

recombinant, expressed in E. coli, >95% (SDS-PAGE)

Synonym(s):

Inflammatory profilin, T Cell Membrane Protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$381.00

$381.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$381.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$381.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

Toxoplasma gondii

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~20 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Toxoplasma gondii ... PRF(7900190)

Biochem/physiol Actions

Profilin is a protozoan parasite protein. It activates toll-like receptor 11 (TLR11), which is an intracellular receptor localized on the endoplasmic reticulum (ER). TLR11 acts as an innate immune sensor for T. gondii profilin (TgPRF).The 12 membrane-spanning ER-resident protein UNC93B1 directly binds TLR11 and control the activation of dendritic cells in response to profilin (Toxoplasma gondii) and parasitic infection in vivo.[1] TLR12 also has an ability to interact with (TgPRF). TLR12 is adequate for recognizing profilin in plasmacytoid dendritic cells (pDCs), whereas TLR11 and TLR12 are both needed in macrophages and conventional DCs. In contrast to TLR11, TLR12-deficient mice fail to resist T. gondii infection.[2]

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Recombinant profilin (Toxoplasma gondii) (aa 1-163).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UNC93B1 is essential for TLR11 activation and IL-12 dependent host resistance to Toxoplasma Gondii.
Pifer R, et al.
The Journal of Biological Chemistry, M110-M110 (2010)
Recognition of profilin by Toll-like receptor 12 is critical for host resistance to Toxoplasma gondii.
Koblansky A A, et al.
Immunity, 38(1), 119-130 (2013)
Erik W Martin et al.
Science signaling, 13(620) (2020-02-27)
The complex signaling dynamics of transcription factors can encode both qualitative and quantitative information about the extracellular environment, which increases the information transfer capacity and potentially supports accurate cellular decision-making. An important question is how these signaling dynamics patterns are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service