Skip to Content
Merck
All Photos(1)

Key Documents

SRP0128

Sigma-Aldrich

DNMT3a Active human

recombinant, expressed in baculovirus infected insect cells

Synonym(s):

DNA (cytosine-5-)-methyltransferase 3A, DNA MTase HsaIIIA, DNMT3A2, M.HsaIIIA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$859.00

$859.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$859.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$859.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

form

aqueous solution

mol wt

128 kDa

packaging

pkg of 10 μg

concentration

>0.02 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DNMT3A(1788)

General description

DNMT3A (DNA methyltransferase 3α) is one of three (DNMT1, DNMT3a and DNMT3b) active forms of DNA methyltransferases.[1] This gene is localized to human chromosome 2p23.[2] DNMT3A is one of the three members of the DNMT3A family, of which DNMT3A and DNMT3B are catalytically active and DNMT3L is an inactive paralogue, which has regulatory actions in germ cells.[3]

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

DNA methyltransferases are responsible for the addition of a methyl residue to the cytosine group of CpG dinucleotides.[4] DNMT3A (DNA methyltransferase 3α) participates in the gene expression suppression by interacting with histone modifiers, such as polycomb-group (PcG). The interaction between these two proteins is thought to be implicated in cancer pathogenesis.[5] During embryogenesis this enzyme is responsible for the formation of genomic methylation patterns and germ line development. Polymorphisms in this gene might serve as biomarkers to determine the prognosis of gastric cancer patients.[1] Mutations in this gene are linked with Tatton-Brown-Rahman syndrome, an overgrowth syndrome characterized by unique facial appearance, intellectual disability, and increased height.[2]

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20 and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatton-Brown-Rahman syndrome due to 2p23 microdeletion.
Okamoto N et al
American Journal of Medical Genetics. Part A, 170A(5), 1339-1342 (2016)
DNMT3A mutations in acute myeloid leukemia.
Ley TJ et al
The New England Journal of Medicine, 363(25), 2424-2433 (2010)
DNMT3A R882 mutants interact with polycomb proteins to block haematopoietic stem and leukaemic cell differentiation.
Koya J et al
Nature Communications, 7, 10924-10924 (2016)
DNA methyltransferase 3a rs1550117 genetic polymorphism predicts poor survival in gastric cancer patients.
Wang C et al
International Journal of Clinical and Experimental Pathology, 8(11), 14864-14874 (2015)
Structure of Dnmt3a bound to Dnmt3L suggests a model for de novo DNA methylation.
Jia D et al
Nature, 449(7159), 248-251 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service