Skip to Content
Merck
All Photos(1)

Key Documents

SML0385

Sigma-Aldrich

ML218

≥98% (HPLC)

Synonym(s):

3,5-Dichloro-N-[[(1α,5α,6-exo,6α)-3-(3,3-dimethylbutyl)-3-azabicyclo[3.1.0]hex-6-yl]methyl]benzamide, CID 45115620, VU0413807, VU0424199-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$168.00

About This Item

Empirical Formula (Hill Notation):
C19H26Cl2N2O
CAS Number:
Molecular Weight:
369.33
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CC(C)(C)CCN1C[C@H]2C(CNC(=O)c3cc(Cl)cc(Cl)c3)[C@H]2C1

InChI

1S/C19H26Cl2N2O/c1-19(2,3)4-5-23-10-16-15(17(16)11-23)9-22-18(24)12-6-13(20)8-14(21)7-12/h6-8,15-17H,4-5,9-11H2,1-3H3,(H,22,24)/t15-,16-,17+

InChI key

GSJIGYLGKSBYBC-OSYLJGHBSA-N

Application

ML218 has been used as a:
  • T-type channel blocker to test its effect on the nociceptive behavior associated with homocysteinemia rats[1]
  • T-type voltage-sensitive calcium channel (VSCC) inhibitor in osteoblasts[2]
  • T-type calcium channel blocker to test its effect on the viability of neural progenitor cells[3]

Biochem/physiol Actions

ML218 (CID 45115620) is a potent and selective T-Type (Cav3.1, Cav3.2, Cav3.3) calcium channel inhibitor (Cav3.2, IC50 = 150 nM in Ca2+ flux; Cav3.2 IC50 = 310 nM and Cav3.3 IC50 = 270 nM with good Drug metabolism/Pharmacokinetics. In a panel of 68 GPCRs, ion channels and transporters, ML218 was found to bind significantly only two other targets (sodium channel site 2 and sigma 1) and had no significant inhibition of L- or N-type calcium channels, KATP or hERG potassium channels. It showed robust inhibition of calcium current in STN neurons and was orally active in a rodent model of Parkinson′s Disease.
ML218 is a potent, selective T-Type calcium channel inhibitor.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brittany D Spitznagel et al.
ACS chemical neuroscience, 11(21), 3658-3671 (2020-11-05)
Malignant migrating partial seizures of infancy is a rare, devastating form of epilepsy most commonly associated with gain-of-function mutations in the potassium channel, Slack. Not only is this condition almost completely pharmacoresistant, there are not even selective drug-like tools available
Andreas Feigenspan et al.
Experimental eye research, 195, 108028-108028 (2020-04-12)
Expression patterns of voltage-gated ion channels determine the spatio-temporal dynamics of ion currents that supply excitable neurons in developing tissue with proper electrophysiological properties. The purpose of the study was to identify fast cationic inward currents in mouse retinal horizontal
Retsu Mitsui et al.
Pflugers Archiv : European journal of physiology, 468(2), 279-291 (2015-11-05)
Postcapillary venules (PCVs) play a critical role in regulating capillary hydrostatic pressure, but their contractile mechanisms are not well understood. We examined the properties of spontaneous vasomotion and corresponding Ca(2+) transients in gastric PCV. In the rat gastric submucosa, changes
Kaori Sato-Numata et al.
The journal of physiological sciences : JPS, 70(1), 49-49 (2020-10-17)
Arginine vasopressin (AVP) neurons play essential roles in sensing the change in systemic osmolarity and regulating AVP release from their neuronal terminals to maintain the plasma osmolarity. AVP exocytosis depends on the Ca2+ entry via voltage-gated Ca2+ channels (VGCCs) in
Ji-Woon Kim et al.
Biomolecules & therapeutics, 26(5), 439-445 (2018-02-22)
T-type calcium channels are low voltage-activated calcium channels that evoke small and transient calcium currents. Recently, T-type calcium channels have been implicated in neurodevelopmental disorders such as autism spectrum disorder and neural tube defects. However, their function during embryonic development

Articles

We offers many products related to calcium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service