Skip to Content
Merck
All Photos(1)

Key Documents

SML4017

Sigma-Aldrich

DiFMU

≥98% (HPLC)

Synonym(s):

6,8-Difluoro-7-hydroxy-4-methyl-2H-1-benzopyran-2-one, 6,8-Difluoro-7-hydroxy-4-methylcoumarin, Marina Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F2O3
CAS Number:
Molecular Weight:
212.15
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=C2C(C(C)=CC(O2)=O)=CC(F)=C1O

InChI key

LLENVBUPWUQAGL-UHFFFAOYSA-N

Biochem/physiol Actions

Fluorescent product of hydrolysis of fluorogenic phosphatase substrate DiFMUP.



DiFMU is a fluorescent product of hydrolysis of fluorogenic phosphatase substrate DiFMUP. DiFMU excitation wavelength is 358nm and emission can be detected at 455 nm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Ge et al.
Analytical biochemistry, 362(1), 63-68 (2007-01-24)
We describe a fluorescence-based assay for the analysis of xylanase activity using a novel fluorogenic substrate, 6,8-difluoro-4-methylumbelliferyl beta-D-xylobioside (DiFMUX(2)). Generation of fluorescent 6,8-difluoro-4-methylumbelliferone (DiFMU) from the substrate corresponded directly to xylanase activity. High-performance liquid chromatography analysis of the digestion products
Lei Sun et al.
Biomolecules, 5(3), 1284-1301 (2015-07-02)
Protein phosphatase 2A (PP2A) is one of the most abundant intracellular serine/threonine (Ser/Thr) phosphatases accounting for 1% of the total cellular protein content. PP2A is comprised of a heterodimeric core enzyme and a substrate-specific regulatory subunit. Potentially, at least seventy
Stefan Welte et al.
Analytical biochemistry, 338(1), 32-38 (2005-02-15)
The fluorogenic substrate 6,8-difluoro-4-methylumbiliferyl phosphate (DIFMUP) has been widely used for the detection of serine and threonine phosphatase activities. Here we describe the use of this substrate for the characterization of protein tyrosine phosphatases (PTPs) and for the screening for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service