Skip to Content
Merck
All Photos(1)

Key Documents

P9233

Sigma-Aldrich

Piribedil maleate salt

Synonym(s):

2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4O6
CAS Number:
Molecular Weight:
414.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

solubility

DMSO: >10 mg/mL
H2O: >10 mg/mL

originator

Servier

storage temp.

2-8°C

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.C1Oc2ccc(CN3CCN(CC3)c4ncccn4)cc2O1

InChI

1S/C16H18N4O2.C4H4O4/c1-4-17-16(18-5-1)20-8-6-19(7-9-20)11-13-2-3-14-15(10-13)22-12-21-14;5-3(6)1-2-4(7)8/h1-5,10H,6-9,11-12H2;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DKGOFBARSQSLOW-BTJKTKAUSA-N

Gene Information

human ... DRD3(1814)

Biochem/physiol Actions

A direct dopamine agonist; may be selective for the D3 subtype; no significant affinity for D1 receptors.

Features and Benefits

This compound was developed by Servier. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Cagnotto et al.
European journal of pharmacology, 313(1-2), 63-67 (1996-10-10)
Receptor binding autoradiography, using the selective ligand [3H]7-OH-(R)DPAT (R(+)-2-dipropylamino-7-hydroxy 1,2,3,4-tetrahydronaphthalene), showed that piribedil is a potent inhibitor at dopamine D3 receptors in limbic regions (island of Calleja), with affinity (IC50) between 30 and 60 nM. The in vitro IC50 of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service