Skip to Content
Merck
All Photos(1)

Key Documents

SML2821

Sigma-Aldrich

DiFMUP

≥98% (HPLC)

Synonym(s):

6,8-Difluoro-4-methylumbelliferyl phosphate, 6,8-Difluoro-4-methyl-7-(phosphonooxy)-2H-1-benzopyran-2-one, 6,8-Difluoro-4-methyl-umbelliferyl phosphate, 6,8-Difluoro-4-methyl-umbelliferyl phosphate, 6,8-Difluoro-7-hydroxy-4-methylcoumarin phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$183.00
25 MG
$736.00

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$183.00
25 MG
$736.00

About This Item

Empirical Formula (Hill Notation):
C10H7F2O6P
CAS Number:
Molecular Weight:
292.13
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CC(=O)Oc2c(F)c(OP(O)(O)=O)c(F)cc12

InChI

1S/C10H7F2O6P/c1-4-2-7(13)17-9-5(4)3-6(11)10(8(9)12)18-19(14,15)16/h2-3H,1H3,(H2,14,15,16)

InChI key

DZANYXOTJVLAEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DiFMUP (6,8-difluoro-4-methylumbelliferyl phosphate), a fluorinated analog of MUP, is a fluorescent synthetic substrate for continuous assessment of phosphatase activity. DiFMUP excitation/emission maxima are 358/450 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon S McCullough et al.
Biochemistry, 57(18), 2584-2589 (2018-04-10)
Although the importance of protein histidine phosphorylation in mammals has been a subject of increasing interest, few chemical probes are available for monitoring and manipulating PHP activity. Here, we present an optimized and validated protocol for assaying the activity of
John S Lazo et al.
The Journal of pharmacology and experimental therapeutics, 371(3), 652-662 (2019-10-12)
Oncogenic protein tyrosine phosphatases (PTPs) are overexpressed in numerous human cancers but they have been challenging pharmacological targets. The emblematic oncogenic PTP4A tyrosine phosphatase family regulates many fundamental malignant processes. 7-Imino-2-phenylthieno[3,2-c]pyridine-4,6(5H,7H)-dione (JMS-053) is a novel, potent, and selective PTP4A inhibitor
Stefan Welte et al.
Analytical biochemistry, 338(1), 32-38 (2005-02-15)
The fluorogenic substrate 6,8-difluoro-4-methylumbiliferyl phosphate (DIFMUP) has been widely used for the detection of serine and threonine phosphatase activities. Here we describe the use of this substrate for the characterization of protein tyrosine phosphatases (PTPs) and for the screening for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service