Skip to Content
Merck
All Photos(1)

Key Documents

SML3071

Sigma-Aldrich

RG3039

≥98% (HPLC)

Synonym(s):

5-[[1-(2,6-Dichlorobenzyl)piperidin-4-yl]methoxy]quinazoline-2,4-diamine, 5-[[1-[(2,6-Dichlorophenyl)methyl]-4-piperidinyl]methoxy]-2,4-quinazolinediamine, D157495, PF 06687859, PF-06687859, PF06687859, RG 3039, RG-3039

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$94.10
25 MG
$382.00

$94.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$94.10
25 MG
$382.00

About This Item

Empirical Formula (Hill Notation):
C21H23Cl2N5O
CAS Number:
Molecular Weight:
432.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$94.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(N)=C2C(OCC3CCN(CC3)CC4=C(C=CC=C4Cl)Cl)=CC=CC2=N1

InChI

1S/C21H23Cl2N5O/c22-15-3-1-4-16(23)14(15)11-28-9-7-13(8-10-28)12-29-18-6-2-5-17-19(18)20(24)27-21(25)26-17/h1-6,13H,7-12H2,(H4,24,25,26,27)

InChI key

MNLHFGXIUJNDAF-UHFFFAOYSA-N

Biochem/physiol Actions

Brain-penetrant, potent mRNA-decapping enzyme DcpS (m7GpppX diphosphatase) inhibitor within in vivo efficacy in a murine spinal muscular atrophy (SMA) model.
RG3039 is a brain-penetrant and potent inhibitor against the scavenger mRNA-decapping enzyme DcpS (m7GpppX diphosphatase) in vitro (mouse DcpS IC50 = 3.4 nM) and in mice in vivo (∼90% & ∼80% inhibition of brain DcpS, respectively, 2 h & 72 h post last 3 mg/kg daily ip. from P1 to P10). In a murine severe spinal muscular atrophy (SMA) model, RG3039 (10-20 mg/kg/d ip. from P1 till death) increases SMAΔ7 mice survival (by 26%) and motor function with a ∼50% increase of VGLUT1 synapses number on L3-L5 motor neurons (33.7/WT mice at P13 vs. 17.2/SMA mice without 26.1/SMA mice with RG3039 treatment).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyssa N Calder et al.
Journal of medicinal chemistry, 59(22), 10067-10083 (2016-08-05)
Spinal muscular atrophy (SMA) is an autosomal recessive neurodegenerative disease resulting from pathologically low levels of survival motor neuron (SMN) protein. The majority of mRNA from the SMN2 allele undergoes alternative splicing and excludes critical codons, causing an SMN protein
Takuji Yamauchi et al.
Cancer cell, 33(3), 386-400 (2018-02-27)
To identify novel targets for acute myeloid leukemia (AML) therapy, we performed genome-wide CRISPR-Cas9 screening using AML cell lines, followed by a second screen in vivo. Here, we show that the mRNA decapping enzyme scavenger (DCPS) gene is essential for AML
Rocky G Gogliotti et al.
Human molecular genetics, 22(20), 4084-4101 (2013-06-06)
Spinal muscular atrophy (SMA) is caused by insufficient levels of the survival motor neuron (SMN) protein due to the functional loss of the SMN1 gene and the inability of its paralog, SMN2, to fully compensate due to reduced exon 7
Blazej A Wojtczak et al.
Journal of the American Chemical Society, 140(18), 5987-5999 (2018-04-21)
The 5' cap consists of 7-methylguanosine (m7G) linked by a 5'-5'-triphosphate bridge to messenger RNA (mRNA) and acts as the master regulator of mRNA turnover and translation initiation in eukaryotes. Cap analogues that influence mRNA translation and turnover (either as
Ariamala Gopalsamy et al.
Journal of medicinal chemistry, 60(7), 3094-3108 (2017-03-04)
The C-5 substituted 2,4-diaminoquinazoline RG3039 (compound 1), a member of a chemical series that was identified and optimized using an SMN2 promoter screen, prolongs survival and improves motor function in a mouse model of spinal muscular atrophy (SMA). It is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service