Skip to Content
Merck
All Photos(1)

Key Documents

SMB01061

Sigma-Aldrich

Emetine

≥90% (LC/MS-ELSD)

Synonym(s):

(-)-Emetine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$253.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C29H40N2O4
CAS Number:
Molecular Weight:
480.64
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

480.64

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

N21[C@@H](C[C@@H]([C@H](C2)CC)C[C@H]4NCCc5c4cc(c(c5)OC)OC)c3c(cc(c(c3)OC)OC)CC1

InChI

1S/C29H40N2O4/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3/t18-,21-,24+,25-/m0/s1

InChI key

AUVVAXYIELKVAI-CKBKHPSWSA-N

General description

Emetine, a pyridoisoquinoline alkaloid, is a bioactive natural compound commonly derived from plants such as Helichrysum revolutum, Sphaerophysa salsula, Coryphantha elephantidens, Uragoga ipecacuanha, Uragoga acuminata and Clonostachys rosea. Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including antiprotozoal, antitumor, anti-inflammatory, anti-infective, antimalarial, and antiviral properties.

Application

Emetine is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and drug discovery research.

Biochem/physiol Actions

Emetine is the principal pyridoisoquinoline of ipecac, from the ground roots of Uragoga (or Cephaelis) ipecacuanha or U. acuminata, of the Rubiaceae.
Emetine is used as an amebicide in many different preparations and may cause serious cardiac, hepatic, or renal damage and violent diarrhea and vomiting. Emetine inhibits protein synthesis in eukaryotic cells but not prokaryotic cells.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service