Skip to Content
Merck
All Photos(1)

Key Documents

SML0294

Sigma-Aldrich

ABL127

≥98% (HPLC)

Synonym(s):

(3R)-1,2-Dicarboxylic acid, 3-cyclopentyl-4-oxo-3-phenyl-1,2-diazetidine 1,2-dimethyl ester, (3R)-Dimethyl 3-cyclopentyl-4-oxo-3-phenyl-1,2-diazetidine-1,2-dicarboxylate, CID24856225

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$195.00
25 MG
$792.00

$195.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$195.00
25 MG
$792.00

About This Item

Empirical Formula (Hill Notation):
C17H20N2O5
CAS Number:
Molecular Weight:
332.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$195.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

film

color

colorless

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

O=C1[C@](C2=CC=CC=C2)(C3CCCC3)N(C(OC)=O)N1C(OC)=O

InChI

1S/C17H20N2O5/c1-23-15(21)18-14(20)17(13-10-6-7-11-13,19(18)16(22)24-2)12-8-4-3-5-9-12/h3-5,8-9,13H,6-7,10-11H2,1-2H3/t17-/m0/s1

InChI key

ZRHWCAFAIHTQKD-KRWDZBQOSA-N

Biochem/physiol Actions

ABL127 is a potent and selective inhibitor of Protein phosphatase methylesterase-1 (PME-1 or PPME-1).
ABL127 is a potent and selective inhibitor of Protein phosphatase methylesterase-1 (PME-1 or PPME-1). IC50 values were 11.1 nM and 6.4 nM in MDA-MB-231 and HEK 293T cells with no activity detected against >50 other serine hydrolases. PME-1 regulates the methylesterification state of protein phosphatase 2A (PP2A) and is implicated in cancer and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunta Ikeda et al.
Journal of biochemistry, 168(6), 643-650 (2020-07-15)
Bone marrow-derived mesenchymal stem cells (BM-MSCs) are multipotent stem cells with ability to self-replicate and differentiate into mesodermal derivatives, such as adipocytes and osteoblasts. BM-MSCs are a critical component of the tumour microenvironment. They support tumour progression by recruiting additional
Sydney A Labuzan et al.
Gene, 739, 144515-144515 (2020-03-01)
Protein phosphatase methylesterase 1 has been identified as a novel gene in skeletal muscle that is upregulated in response to neurogenic atrophy in mice. Western blot analysis confirms that Ppme1 is expressed during both muscle cell proliferation and differentiation. Additionally
Bin Wu et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 17(4), 1878-1896 (2020-09-23)
The molecular mechanism of Alzheimer-like cognitive impairment induced by manganese (Mn) exposure has not yet been fully clarified, and there are currently no effective interventions to treat neurodegenerative lesions related to manganism. Protein phosphatase 2 A (PP2A) is a major
Ryotaro Yabe et al.
FEBS open bio, 8(9), 1486-1496 (2018-09-07)
Reversible methyl-esterification (methylation) of Leu309 in the protein phosphatase 2A catalytic subunit (PP2Ac) is essential for proper biogenesis of the PP2A holoenzyme. Accumulating evidence links PP2Ac methylation to diseases, including cancer and neurodegenerative disorders. Protein phosphatase methyl-esterase (PME-1) specifically catalyzes
Ling Meng et al.
Frontiers in immunology, 12, 648913-648913 (2021-04-30)
The excessive M1 polarization of macrophages drives the occurrence and development of inflammatory diseases. The reprogramming of macrophages from M1 to M2 can be achieved by targeting metabolic events. Taurine promotes for the balance of energy metabolism and the repair

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service