Skip to Content
Merck
All Photos(1)

Key Documents

SML3010

Sigma-Aldrich

Belinostat

≥98% (HPLC)

Synonym(s):

(2E)-N-Hydroxy-3-[3-(phenylsulfamoyl)phenyl]prop-2-enamide, (2E)-N-Hydroxy-3-[3-[(phenylamino)sulfonyl]phenyl]-2-propenamide, (E)-N-Hydroxy-3-(3-(N-phenylsulfamoyl)phenyl)acrylamide, PX 105684, PX-105684, PX105684, PXD 101, PXD-101, PXD101

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$119.00
50 MG
$482.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$119.00
50 MG
$482.00

About This Item

Empirical Formula (Hill Notation):
C15H14N2O4S
CAS Number:
Molecular Weight:
318.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(Nc2ccccc2)c1cc(ccc1)\C=C\C(=O)NO

InChI

1S/C15H14N2O4S/c18-15(16-19)10-9-12-5-4-8-14(11-12)22(20,21)17-13-6-2-1-3-7-13/h1-11,17,19H,(H,16,18)/b10-9+

InChI key

NCNRHFGMJRPRSK-MDZDMXLPSA-N

Biochem/physiol Actions

Belinostat (PX105684; PXD101) is a potent, hydroxamate class pan-selective histone deacetylases (HDACs) inhibitor (IC50: class I HDAC1/2/3/8 = 41/125/30/216 nM, class IIA HDAC4/7/9 = 115/67/128 nM, class IIB HDAC6 = 82 nM) with anti-cancer efficacy in vitro and in vivo.
Potent, hydroxamate class pan-selective histone deacetylases (HDACs) inhibitor with anti-cancer efficacy in vitro and in vivo.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service