Skip to Content
Merck
All Photos(2)

Key Documents

840465P

Avanti

18:0 PG

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioctadecanoyl-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); DSPG; PG(18:0/18:0); 110649

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$130.00
200 MG
$375.00
1 G
$965.00

$130.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$130.00
200 MG
$375.00
1 G
$965.00

About This Item

Empirical Formula (Hill Notation):
C42H82O10PNa
CAS Number:
Molecular Weight:
801.06
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$130.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

1,2-distearoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840465P-1g)
pkg of 1 × 200 mg (840465P-200mg)
pkg of 1 × 25 mg (840465P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C42H83O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h39-40,43-44H,3-38H2,1-2H3,(H,47,48);/q;+1/p-1/t39?,40-;/m1./s1

InChI key

YNQYZBDRJZVSJE-QTOMIGAPSA-M

Packaging

20 mL Clear Glass Screw Cap Vial (840465P-1g)
5 mL Amber Glass Screw Cap Vial (840465P-200mg)
5 mL Amber Glass Screw Cap Vial (840465P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brenda L Kessenich et al.
ACS omega, 5(21), 12154-12161 (2020-06-18)
Amphiphilic molecules can alter the wettability of soil minerals. To determine how the headgroup chemistry of amphiphiles determines these effects, we investigate a system of the clay montmorillonite with long-chain phospholipids. We use phosphatidylglycerol (PG) phospholipids to contrast with our
Lavanya Visvabharathy et al.
PLoS pathogens, 16(4), e1008443-e1008443 (2020-04-29)
Staphylococcus aureus (SA) is the causative agent of both skin/soft tissue infections as well as invasive bloodstream infections. Though vaccines have been developed to target both humoral and T cell-mediated immune responses against SA, they have largely failed due to
Margaret W Ndinguri et al.
Journal of drug delivery, 2012, 592602-592602 (2012-12-06)
Cancer treatment by chemotherapy is typically accompanied by deleterious side effects, attributed to the toxic action of chemotherapeutics on proliferating cells from nontumor tissues. The cell surface proteoglycan CD44 has been recognized as a cancer stem cell marker. The present
N Basaran Mutlu-Agardan et al.
Journal of liposome research, 1-12 (2020-04-16)
Amphotericin B (AmB) is a very potent antibiotic which still remains as the gold standard for the treatment of systemic fungal infections. AmB is a member of Biopharmaceutical Classification System Class IV, mainly characterized by its poor solubility and low
Parvin Zamani et al.
Life sciences, 252, 117646-117646 (2020-04-10)
Myeloid-derived suppressor cells (MDSCs) are immunosuppressive cells causing resistance to immunotherapies in cancer tumors. In the current study, various immunogenic and therapeutic features of the combination therapies with non-liposomal Doxorubicin (Dox) and the E75 immunogenic peptide (Pep), derived from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service