Skip to Content
Merck
All Photos(1)

Key Documents

SML2794

Sigma-Aldrich

Erastin2

≥98% (HPLC)

Synonym(s):

2-((4-(2-(4-Chlorophenoxy)acetyl)piperazin-1-yl)methyl)-3-(4-isopropoxybiphenyl-3-yl)quinazolin-4(3H)-one, 2-[[4-[2-(4-Chlorophenoxy)acetyl]-1-piperazinyl]methyl]-3-[4-(1-methylethoxy)[1,1′-biphenyl]-3-yl]-4(3H)-quinazolinone, 35MEW28

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$334.00
25 MG
$515.00

$334.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$334.00
25 MG
$515.00

About This Item

Empirical Formula (Hill Notation):
C36H35ClN4O4
CAS Number:
Molecular Weight:
623.14
UNSPSC Code:
12352200
NACRES:
NA.77

$334.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N(C1=C(OC(C)C)C=CC(C2=CC=CC=C2)=C1)C(CN3CCN(C(COC4=CC=C(Cl)C=C4)=O)CC3)=N5)C6=C5C=CC=C6

Biochem/physiol Actions

Erastin analog with greatly improved system xc- (SLC7A11/xCT + SLC3A2/4F2hc) inhibitory potency and ferropotosis induction efficacy
Erastin2 is an Erastin analog with greatly improved system xc- (SLC7A11/xCT + SLC3A2/4F2hc) inhibitory potency (Erastin2 IC50 = 3.5 nM/CCF-STTG1; Erastin IC50 = 200 nM/HT-1080, 140 nM/Calu-1; Glu release assay) and ferropotosis induction efficacy (HT-1080 EC50 = 150 nM/Erastin2 vs. 1.2 μM/Erastin).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allegra T Aron et al.
Journal of the American Chemical Society, 138(43), 14338-14346 (2016-11-03)
Iron is essential for sustaining life, as its ability to cycle between multiple oxidation states is critical for catalyzing chemical transformations in biological systems. However, without proper regulation, this same redox capacity can trigger oxidative stress events that contribute to
Amy Tarangelo et al.
Cell reports, 22(3), 569-575 (2018-01-19)
How cancer cells respond to nutrient deprivation remains poorly understood. In certain cancer cells, deprivation of cystine induces a non-apoptotic, iron-dependent form of cell death termed ferroptosis. Recent evidence suggests that ferroptosis sensitivity may be modulated by the stress-responsive transcription
Mariana Figuera-Losada et al.
Biochemistry and biophysics reports, 9, 266-272 (2017-09-29)
The inflammatory response in the central nervous system involves activated microglia. Under normal conditions they remove damaged neurons by phagocytosis. On the other hand, neurodegenerative diseases are thought to involve chronic microglia activation resulting in release of excess glutamate, proinflammatory
Bei Liu et al.
Biochemical and biophysical research communications, 497(1), 233-240 (2018-02-11)
Heart failure (HF) is the end stage of cardiovascular disease and is characterized by the loss of myocytes caused by cell death. Puerarin has been found to improve HF clinically, and animal study findings have confirmed its anti-cell-death properties. However
Giovanni C Forcina et al.
Cell systems, 4(6), 600-610 (2017-06-12)
Cytotoxic compounds are important drugs and research tools. Here, we introduce a method, scalable time-lapse analysis of cell death kinetics (STACK), to quantify the kinetics of compound-induced cell death in mammalian cells at the population level. STACK uses live and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service