Skip to Content
Merck
All Photos(1)

Key Documents

SML2974

Sigma-Aldrich

Apilimod

≥98% (HPLC)

Synonym(s):

3-Methyl-benzaldehyde 2-[6-(4-morpholinyl)-2-[2-(2-pyridinyl)ethoxy]-4-pyrimidinyl]hydrazone, STA 5326, STA-5326

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$544.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C23H26N6O2
CAS Number:
Molecular Weight:
418.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N4(CCOCC4)c1nc(nc(c1)N\N=C\c3cc(ccc3)C)OCCc2ncccc2

InChI

1S/C23H26N6O2/c1-18-5-4-6-19(15-18)17-25-28-21-16-22(29-10-13-30-14-11-29)27-23(26-21)31-12-8-20-7-2-3-9-24-20/h2-7,9,15-17H,8,10-14H2,1H3,(H,26,27,28)/b25-17+

InChI key

HSKAZIJJKRAJAV-KOEQRZSOSA-N

Biochem/physiol Actions

Apilimod (STA-5326) is a potent, orally active and selective inhibitor of the cytokines IL-12 and IL-23. Apilimod selectively suppresses synthesis of IL-12 and IL-23 by inhibition of phosphotransferase activity of TLR-4 regulator lipid kinase PIKfyve (phosphatidylinositol-3-phosphate 5-kinase). It suppresses Th1 cells response in vivo and reduces inflammatory histopathologic changes. Also, it exhibits strong anti COVID-19 activity in two cell lines (Vero E6:IC50 < 0.08 μM; A549-ACE2:IC50 = 0.007 μM). Apilimod potently reduces entry of SARS-CoV S pseudovirions on 293/hACE2 cells.
potent, orally active and selective IL-12 and IL-23 inhibitor; PIKfyve inhibitor; Exhibit strong anti COVID-19 activity in cell lines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinming Cai et al.
Chemistry & biology, 20(7), 912-921 (2013-07-31)
Toll-like receptor (TLR) signaling is a key component of innate immunity. Aberrant TLR activation leads to immune disorders via dysregulation of cytokine production, such as IL-12/IL-23. Herein, we identify and characterize PIKfyve, a lipid kinase, as a critical player in
Xiuyuan Ou et al.
Nature communications, 11(1), 1620-1620 (2020-03-30)
Since 2002, beta coronaviruses (CoV) have caused three zoonotic outbreaks, SARS-CoV in 2002-2003, MERS-CoV in 2012, and the newly emerged SARS-CoV-2 in late 2019. However, little is currently known about the biology of SARS-CoV-2. Here, using SARS-CoV-2 S protein pseudovirus
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service