Skip to Content
Merck
All Photos(1)

Key Documents

SML1504

Sigma-Aldrich

Tegaserod maleate

≥98% (HPLC)

Synonym(s):

2-[(5-Methoxy-1H-indol-3-yl)methylene]-N-pentyl-hydrazinecarboximidamide maleate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$151.00
50 MG
$616.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$151.00
50 MG
$616.00

About This Item

Empirical Formula (Hill Notation):
C16H23N5O · C4H4O4
CAS Number:
Molecular Weight:
417.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N=C(NCCCCC)N/N=C/C1=CNC2=C1C=C(OC)C=C2.OC(/C=C\C(O)=O)=O

InChI

1S/C16H23N5O.C4H4O4/c1-3-4-5-8-18-16(17)21-20-11-12-10-19-15-7-6-13(22-2)9-14(12)15;5-3(6)1-2-4(7)8/h6-7,9-11,19H,3-5,8H2,1-2H3,(H3,17,18,21);1-2H,(H,5,6)(H,7,8)/b20-11+;2-1-

InChI key

CPDDZSSEAVLMRY-FEQFWAPWSA-N

Gene Information

human ... HTR4(3360)

Application

Tegaserod maleate has been used to study its cytotoxic effects on human astrocytes and to study its effects as an anti-glioblastoma agent.[1]

Biochem/physiol Actions

It stimulates neurotransmitter release from enteric nerves and regularizes gastrointestinal function. Tegaserod has been used to treat irritable bowel syndrome associated with constipation and chronic constipation.[2]
Tegaserod is a potent full agonist at serotonin 5-HT4 receptors with an EC50 of 7 nM.
Tegaserod is a serotonin 5-HT4 agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening of clinically-applicable drugs in patient-derived glioblastoma stem-like cells identifies amiodarone, clofazimine and triptolide as potential anti-glioma agents
Sigrid S
Combinatorial Chemistry: From Theory to Application (2015)
Corrado Blandizzi and Carmelo Scarpignato
Side Effects of Drugs Annual: A world-wide yearly survey of new data and trends in adverse drug reactions., 33, 741-767 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service