Skip to Content
Merck
All Photos(1)

Key Documents

SML2635

Sigma-Aldrich

Z944 hydrochloride

≥98% (HPLC)

Synonym(s):

4-[[(3-Chloro-5-fluorobenzoyl)amino]methyl]-N-(1,1-dimethylethyl)-1-piperidineacetamide hydrochloride, N-((1-(2-(tert-Butylamino)-2-oxoethyl)piperidin-4-yl)methyl)-3-chloro-5-fluorobenzamide hydrochloride, Z 944 hydrochloride, Z-944

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$500.00

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$124.00
25 MG
$500.00

About This Item

Empirical Formula (Hill Notation):
C19H27ClFN3O2 · xHCl
Molecular Weight:
383.89 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Orally available, state-dependent, potent and selective T-type calcium channel blocker (Cav3.1, Cav3.2, Cav3.3) with in vivo absence seizure-reducing efficacy.
Z944 is an orally available, state-dependent, potent and selective T-type calcium channel blocker (hCav3.1/Cav3.2/Cav3.3 IC50 by voltage clamp = 50/160/110 nM under ~30% inactivated state & 130/540/260 nM in the closed state) over non-T-type voltage-gated channels (rCaV2.2 IC50 = 11 μM/30% inactivated & 150 μM/closed; hERG/rCaV1.2/hNaV1.5 IC50 = 7.8/32/100 μM). Z944 effectively suppresses seizure activity in the GAERS (absence epilepsy rats from Strasbourg) model in vivo (10-30 mg/kg qod ip.) with no adverse cardiovascular effects or neurotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z944: a first in class T-type calcium channel modulator for the treatment of pain.
M Lee
Journal of the peripheral nervous system : JPNS, 19 Suppl 2, S11-S12 (2014-10-02)
Wendie N Marks et al.
Neuroscience, 332, 121-129 (2016-07-02)
The role of T-type calcium channels in brain diseases such as absence epilepsy and neuropathic pain has been studied extensively. However, less is known regarding the involvement of T-type channels in cognition and behavior. Prepulse inhibition (PPI) is a measure
Dongyang Huang et al.
Antioxidants & redox signaling, 25(5), 233-251 (2016-06-17)
Neuropeptide substance P (SP) is produced and released by a subset of peripheral sensory neurons that respond to tissue damage (nociceptors). SP exerts excitatory effects in the central nervous system, but peripheral SP actions are still poorly understood; therefore, here
Isamu Aiba et al.
Journal of neurophysiology, 121(4), 1266-1278 (2019-01-31)
Cholinergic vagal nerves projecting from neurons in the brain stem nucleus ambiguus (NAm) play a predominant role in cardiac parasympathetic pacemaking control. Central adrenergic signaling modulates the tone of this vagal output; however, the exact excitability mechanisms are not fully
Mark T Henbid et al.
Experimental neurology, 296, 16-22 (2017-06-29)
Childhood absence epilepsy (CAE) is associated with interictal co-morbid symptoms including abnormalities in social behaviour. Genetic Absence Epilepsy Rats from Strasbourg (GAERS) is a model of CAE that exhibits physiological and behavioural alterations characteristic of the human disorder. However, it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service