Skip to Content
Merck
All Photos(1)

Key Documents

SML2508

Sigma-Aldrich

Etamsylate

Synonym(s):

Etamsylate, Cyclonamine, ETHAMSYLATE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO5S
CAS Number:
Molecular Weight:
263.31
UNSPSC Code:
12352200
NACRES:
NA.77

form

powder

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])c1c(ccc(c1)O)O.[N+H2](CC)CC

InChI

1S/C6H6O5S.C4H11N/c7-4-1-2-5(8)6(3-4)12(9,10)11;1-3-5-4-2/h1-3,7-8H,(H,9,10,11);5H,3-4H2,1-2H3

InChI key

HBGOLJKPSFNJSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Etamsylate (or ethamsylate) is a haemostatic agent; also promotes angioprotective and proaggregant action. It stimulates thrombopoiesis and their release from bone marrow. Haemostatic action is due to activation of thromboplastin formation on damaged sites of small blood vessels and decrease of PgI2 (Prostacyclin I2) synthesis; it also facilitates platelet aggregation and adhesion, that at last induce decrease and stop of hemorrhage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service