Skip to Content
Merck
All Photos(1)

Key Documents

E1830000

Etamsylate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cyclonamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO5S
CAS Number:
Molecular Weight:
263.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

etamsylate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])c1c(ccc(c1)O)O.[N+H2](CC)CC

InChI

1S/C6H6O5S.C4H11N/c7-4-1-2-5(8)6(3-4)12(9,10)11;1-3-5-4-2/h1-3,7-8H,(H,9,10,11);5H,3-4H2,1-2H3

InChI key

HBGOLJKPSFNJSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etamsylate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rod Hunt et al.
The Cochrane database of systematic reviews, (1)(1), CD004343-CD004343 (2010-01-22)
Ethamsylate decreases blood loss in certain clinical situations such as menorrhagia and following some surgical procedures. This potential to reduce bleeding has led to the hypothesis that it may have a role to play in reducing intraventricular haemorrhage in preterm
Takehito Naruoka et al.
Hinyokika kiyo. Acta urologica Japonica, 54(5), 341-344 (2008-06-13)
We examined the effectiveness of supplemental administration of Eviprostat in patients with benign prostatic hyperplasia (BPH) whose lower urinary tract symptoms (LUTS) caused by BPH were not adequately relieved by an alpha1-adrenoceptor blocker. Twenty-nine patients with insufficient improvement in the
Alaa El-Gindy et al.
Journal of AOAC International, 96(4), 730-736 (2013-09-05)
A simple, selective, sensitive, accurate, and precise method was developed for determination of ethamsylate (ET) in human urine and in ET tablets using RP-HPLC. The method uses a C18 (5 pm particle size) column at ambient temperature with the mobile
Ondrej Wiewiorka et al.
Scandinavian journal of clinical and laboratory investigation, 73(5), 449-451 (2013-05-22)
With discrepancies encountered as early as the verification of enzymatic method for quantification of serum creatinine, our research pointed to a later confirmed interference caused by a compound called ethamsylate present in the commonly used antihemorrhagic drug Dicynone. We measured
Editorial comment to suppression of bladder overactivity and oxidative stress by the phytotherapeutic agent, Eviprostat, in a rat model of atherosclerosis-induced chronic bladder ischemia.
Shizuo Yamada
International journal of urology : official journal of the Japanese Urological Association, 19(7), 675-675 (2012-06-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service