Skip to Content
Merck
All Photos(4)

Key Documents

H0888

Sigma-Aldrich

Hydrocortisone

BioReagent, suitable for cell culture

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein:
1354819
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
form:
powder
Assay:
≥98%

product line

BioReagent

Quality Level

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

211-214 °C (lit.)

solubility

chloroform/ethanol (1:1): 50 mg/mL, clear, colorless to very faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrocortisone has been used as a supplement in culture media for oligodendrocytes, colon cells and epithelial cells.

Biochem/physiol Actions

Hydrocortisone (also referred to as cortisol) is an endogenous glucocorticoid which is secreted by the adrenal gland. It is produced from the hypothalamus–pituitary–adrenal (HPA) axis. Hydrocortisone interacts with glucocorticoid receptors and regulates various signaling pathways. It is associated with the induction of energy mobilization, increase in cerebral perfusion rates and local glucose consumption. It is also linked with increase in cardiovascular output and respiration, redistribution of blood, delivery of molecules and energy to the brain and muscles and modulation of immune responses.
Primary glucocorticoid secreted by the adrenal cortex.
Primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na2+ retention potency.

Physical form

powder-RT; stock-frozen in working aliquots, avoid repeated freeze/thaw

Reconstitution

To prepare 50 μg/mL stock solution, add 1.0 mL absolute ethanol to 1.0 mg of product, gently swirl to dissolve, add 19 mL sterile medium.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Adrenal gland,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunomodulation in septic shock: hydrocortisone differentially regulates cytokine responses.
Briegel J et al.
Journal of the American Society of Nephrology, 12, S70-S70 (2001)
The cortisol awakening response (CAR): facts and future directions.
Fries E et al.
International Journal of Psychophysiology, 72, 67-67 (2009)
Human colon tissue in organ culture: preservation of normal and neoplastic characteristics.
Dame MK et al.
In Vitro Cellular & Developmental Biology. Animal, 46, 114-114 (2010)
Marc Hafner et al.
Cell chemical biology, 26(8), 1067-1080 (2019-06-11)
The target profiles of many drugs are established early in their development and are not systematically revisited at the time of FDA approval. Thus, it is often unclear whether therapeutics with the same nominal targets but different chemical structures are
A J Kent et al.
Biomaterials, 218, 119337-119337 (2019-07-22)
Laminin-111 (Ln-1), an extracellular matrix (ECM) glycoprotein found in the basement membrane of mammary gland epithelia, is essential for lactation. In mammary epithelial cells (MECs), dystroglycan (Dg) is believed to be necessary for polymerization of laminin-111 into networks., thus we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service