Skip to Content
Merck
All Photos(1)

Key Documents

SML1918

Sigma-Aldrich

ML349

≥98% (HPLC)

Synonym(s):

ML349, (5,5-Dioxido-4H-thieno[3,2-c][1]benzothiopyran-2-yl)[4-(4-methoxyphenyl)-1-piperazinyl]methanone, ML 349, ML-349, (5,5-Dioxo-4H-thieno[4,5-c]thiochromen-2-yl)-[4-(4-methoxyphenyl)piperazin-1-yl]methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$532.00

$132.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$532.00

About This Item

Empirical Formula (Hill Notation):
C23H22N2O4S2
CAS Number:
Molecular Weight:
454.56
UNSPSC Code:
12352200
NACRES:
NA.77

$132.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S]1(=O)(=O)Cc2c([s]c(c2)C(=O)N4CCN(CC4)c5ccc(cc5)OC)c3c1cccc3

InChI

1S/C23H22N2O4S2/c1-29-18-8-6-17(7-9-18)24-10-12-25(13-11-24)23(26)20-14-16-15-31(27,28)21-5-3-2-4-19(21)22(16)30-20/h2-9,14H,10-13,15H2,1H3

InChI key

YVIJPELUPZUEJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

ML349 is a substrate-competitive, reversible and APT2-selective acyl-protein thioesterase (APT) inhibitor (IC50/Ki = 510 nM/230 nM against APT2; IC50 & Ki >10 μM against APT1). ML349 effectively inhibits cellular APT2 activity in cultures (by >95% in HEK293T & mouse T cells; 5 μM for 4 h) and in mice in vivo (by >90% in lung/heart/kidney and by ~50% in brain tissue 4 h post 50 mg/kg i.p. dosing) without affecting more than 15 cellular serine hydrolases and APT1. ML349 treatment (5 μM) is shown to restore palmitoylation and membrane localization of the multidomain scaffolding protein Scribble (Scrib) in MDCK cells overexpressing the epithelial-to-mesenchymal transcription factor (EMT-TF) Snail.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Adibekian et al.
Journal of the American Chemical Society, 134(25), 10345-10348 (2012-06-14)
The development of small-molecule inhibitors for perturbing enzyme function requires assays to confirm that the inhibitors interact with their enzymatic targets in vivo. Determining target engagement in vivo can be particularly challenging for poorly characterized enzymes that lack known biomarkers
Rahul S Kathayat et al.
Nature chemical biology, 13(2), 150-152 (2016-12-20)
Hundreds of human proteins are modified by reversible palmitoylation of cysteine residues (S-palmitoylation), but the regulation of depalmitoylation is poorly understood. Here, we develop 'depalmitoylation probes' (DPPs), small-molecule fluorophores, to monitor the endogenous activity levels of 'erasers' of S-palmitoylation, acylprotein
Jeannie L Hernandez et al.
Cell chemical biology, 24(1), 87-97 (2017-01-10)
The multidomain scaffolding protein Scribble (Scrib) organizes key signaling complexes to specify basolateral cell polarity and suppress aberrant growth. In many human cancers, genetically normal Scrib mislocalizes from cell-cell junctions to the cytosol, correlating with enhanced growth signaling and malignancy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service