Skip to Content
Merck
All Photos(1)

Key Documents

925047

Sigma-Aldrich

KB02-COOH

Synonym(s):

2-((1-(2-Chloroacetyl)-1,2,3,4-tetrahydroquinolin-6-yl)oxy)acetic acid, Functionalized scout fragment

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$149.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$149.00

About This Item

Empirical Formula (Hill Notation):
C13H14ClNO4
CAS Number:
Molecular Weight:
283.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$149.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

reaction suitability

reagent type: chemical modification reagent

Quality Level

storage temp.

2-8°C

Application

KB02-COOH is a cysteine-reactive small-molecule fragment for chemoproteomic and ligandability studies for both traditionally druggable proteins as well as ″undruggable,″ or difficult-to-target, proteins. This fragment electrophile is the functionalized version of KB02 (912131).

Related useful products may include cysteine fragment KB02 (912131), KB02-SLF (914738), IA alkyne (924237), IA 5-TAMRA (925020), desthiobiotin iodoacetamide (923826), or biotin iodoacetamide (B2059).

Technology spotlight: Proteomic Ligandability Assessment

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Zhang et al.
Nature chemical biology, 15(7), 737-746 (2019-06-19)
Ligand-dependent protein degradation has emerged as a compelling strategy to pharmacologically control the protein content of cells. So far, however, only a limited number of E3 ligases have been found to support this process. Here, we use a chemical proteomic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service