Skip to Content
Merck
All Photos(1)

Key Documents

SML1901

Sigma-Aldrich

ML348

≥98% (HPLC)

Synonym(s):

ML 348, ML-348, N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-[4-(2-furoyl)-1-piperazinyl]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$532.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$532.00

About This Item

Empirical Formula (Hill Notation):
C18H17ClF3N3O3
CAS Number:
Molecular Weight:
415.79
UNSPSC Code:
51111800
NACRES:
NA.77

$132.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)Cl)NC(=O)CN2CCN(CC2)C(=O)c3[o]ccc3

InChI

1S/C18H17ClF3N3O3/c19-13-4-3-12(18(20,21)22)10-14(13)23-16(26)11-24-5-7-25(8-6-24)17(27)15-2-1-9-28-15/h1-4,9-10H,5-8,11H2,(H,23,26)

InChI key

OXKNHBBDOIMFFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

ML348 is a substrate-competitive, reversible and APT1-selective acyl-protein thioesterase (APT) inhibitor (IC50/Ki = 840 nM/300 nM against APT1; IC50 & Ki >10 μM against APT2). ML348 effectively inhibits cellular APT1 activity in cultures (by >95% in HEK293T & mouse T cells; 5 μM for 4 h) and in mice in vivo (by >90% in lung/heart/kidney and by ~50% in brain tissue 4 h post 50 mg/kg i.p. dosing) without affecting more than 15 cellular serine hydrolases and APT2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeannie L Hernandez et al.
Cell chemical biology, 24(1), 87-97 (2017-01-10)
The multidomain scaffolding protein Scribble (Scrib) organizes key signaling complexes to specify basolateral cell polarity and suppress aberrant growth. In many human cancers, genetically normal Scrib mislocalizes from cell-cell junctions to the cytosol, correlating with enhanced growth signaling and malignancy.
Rahul S Kathayat et al.
Nature chemical biology, 13(2), 150-152 (2016-12-20)
Hundreds of human proteins are modified by reversible palmitoylation of cysteine residues (S-palmitoylation), but the regulation of depalmitoylation is poorly understood. Here, we develop 'depalmitoylation probes' (DPPs), small-molecule fluorophores, to monitor the endogenous activity levels of 'erasers' of S-palmitoylation, acylprotein
Alexander Adibekian et al.
Journal of the American Chemical Society, 134(25), 10345-10348 (2012-06-14)
The development of small-molecule inhibitors for perturbing enzyme function requires assays to confirm that the inhibitors interact with their enzymatic targets in vivo. Determining target engagement in vivo can be particularly challenging for poorly characterized enzymes that lack known biomarkers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service