Skip to Content
Merck
All Photos(6)

Documents

Z744035

Sigma-Aldrich

Penn PhD Photoreactor M2

Synonym(s):

photoreactor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.21

AC/DC input

100 - 240 V AC, 50/60 Hz

feature

thermocouple type K-Type Thermocouple
(Touch Screen: 3.5” TFT LCD; 320 x 480 resolution)

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

parameter

(Variable stir bar control 100 - 2000 RPM)
0-95% RH at 10-40 °C

W × H × D

11.4 cm × 27.2 cm × 27.9 cm
4.5 in. × 10.7 in. × 11.0 in.

shipped in

ambient

General description

The Penn PhD Photoreactor M2 is a benchtop instrument designed for chemists and researchers to accelerate chemical reactions using photoredox catalysis. The Photoreactor M2 combines LED illumination, mechanical stirring and cooling into one device. The user defined parameters of temperature, intensity, stir rate and time, create a valuable tool for repeatability, traceability, efficiency and consistency of results. The Photoreactor M2 addresses the potential to streamline synthetic sequences, and create valuable strategies for addressing some of the challenges of molecule construction in drug discovery.

Features and Benefits

  • Modular design allows for use with a variety of wavelengths: 450 nm (included), 420 nm (sold separately), 395 nm (sold separately) and 365 nm (sold separately)
  • 360 degree reflective environment maximizes surface area photon capture
  • Light shield interlock prevents user exposure to harmful light rays
  • Interactive touch screen controls reaction parameters
  • Intertek ETL, CE, and CB approved
  • User defined parameters including temperature, light intensity, fan speed and stirring
  • Auto stop, pause and reset options
  • Supports vial sizes gc, 4, 8, 20, 40 ml
  • Multi-vial holders are available to allow four 8 mL vials and five 4 mL vials reactions to be run in parallel.
  • Temp feedback using a k-type thermocouple
Regional Plug Types Available
  • Z744035-1EA-US: type B
  • Z744035-1EA- IN: type D
  • Z744035-1EA-EU-E: type E
  • Z744035-1EA-EU-F: type F
  • Z744035-1EA-UK: type G
  • Z744035-1EA-CN: type I
  • Z744035-1EA-CH: type J

Photocatalysis Technology Spotlight

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.

Photoredox catalysis is a powerful synthetic methodology to form challenging covalent bonds using light irradiation. It is effective for light-driven polymer and small molecule synthesis.

Effective Csp2- and Csp-hybridized coupling reactions established for catalysis, expanding to multi-step Csp3-coupling.

The application of radical chemistry towards organic synthesis is well-developed and wide-reaching, though often hampered by a dependence on toxic radical initiators.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service