Skip to Content
Merck
All Photos(1)

Key Documents

SML1839

Sigma-Aldrich

PFM39

≥98% (HPLC)

Synonym(s):

(5Z)-2-Amino-5-[(4-aminophenyl)methylene]-4(5H)-thiazolone, 5-(4-Aminobenzylidene)-2-iminothiazolidin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$161.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$161.00

About This Item

Empirical Formula (Hill Notation):
C10H9N3OS
CAS Number:
Molecular Weight:
219.26
UNSPSC Code:
12352200
NACRES:
NA.77

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=CC=C(/C=C2SC(NC\2=O)=N)C=C1

InChI

1S/C10H9N3OS/c11-7-3-1-6(2-4-7)5-8-9(14)13-10(12)15-8/h1-5H,11H2,(H2,12,13,14)/b8-5-

InChI key

QXOIZYPBCJHYLN-YVMONPNESA-N

Biochem/physiol Actions

PFM39 is a potent cell-permeable Mirin analog that selectively inhibits MRE11 exo-, but not endo-, nuclease activity.
PFM39 is a potent cell-permeable Mirin analog that selectively inhibits MRE11 exo-, but not endo-, nuclease activity. PFM39 targets MRE11 in a fashion similar to Mirin, but distinct from that of PFM01 to allow a blockage of dsDNA phosphate backbone rotation and selective inhibition against MRE11 exo-, but not endo-, nuclease activity. FM39 potently impairs G2-phase double-strand break (DSB) repair in 1BR3-hTERT fibrolasts following ionizing irradiation (IR).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronan Broderick et al.
Nature cell biology, 18(3), 271-280 (2016-01-26)
Repair of DNA double-strand breaks (DSBs) by homologous recombination (HR) is critical for survival and genome stability of individual cells and organisms, but also contributes to the genetic diversity of species. A vital step in HR is MRN-CtIP-dependent end resection
Atsushi Shibata et al.
Molecular cell, 53(1), 7-18 (2013-12-10)
MRE11 within the MRE11-RAD50-NBS1 (MRN) complex acts in DNA double-strand break repair (DSBR), detection, and signaling; yet, how its endo- and exonuclease activities regulate DSBR by nonhomologous end-joining (NHEJ) versus homologous recombination (HR) remains enigmatic. Here, we employed structure-based design
Lea Völkening et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2812-2820 (2020-01-08)
The Mre11A/RAD50/NBN complex (MRN) is an essential regulator of the cellular damage response after DNA double-strand breaks (DSBs). More recent work has indicated that MRN may also impact on the duration of mitosis. We show here that RAD50-deficient fibroblasts exhibit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service