Skip to Content
Merck
All Photos(1)

Key Documents

62970

Sigma-Aldrich

Lysozyme from chicken egg white

dialyzed, lyophilized, powder, ~100000 U/mg

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$97.50
5 G
$339.00

$97.50


Estimated to ship onMarch 27, 2025



Select a Size

Change View
1 G
$97.50
5 G
$339.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$97.50


Estimated to ship onMarch 27, 2025


biological source

chicken egg white

Quality Level

form

powder

quality

dialyzed
lyophilized

specific activity

~100000 U/mg

mol wt

single-chain 14.3 kDa
Mr ~14600

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

UniProt accession no.

application(s)

cell analysis

storage temp.

2-8°C

Gene Information

chicken ... LYZ(396218)

Looking for similar products? Visit Product Comparison Guide

Application

Lysozyme from chicken egg white has been used:
  • as a standard in small-angle neutron scattering experiments [1] and infrared absorbance spectroscopy[2]
  • as a constituent of cell lysis buffer[3]
  • in protein crystallization experiments[4]

Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).

Unit Definition

1 U corresponds to the amount of enzyme which decreases the absorbance at 450 nm by 0.001 per minute at pH 7.0 and 25°C (Micrococcus luteus, ATCC 4698, as substrate)

Other Notes

Suitable for the hydrolysis of bacterial cell walls, mucopolysaccharides, mucopolypeptides or chitin; degradability of L. lactis subsp. cremoris H2 cell walls[5]; Review[6][7][8]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Coolbear et al.
Applied and environmental microbiology, 58(10), 3263-3270 (1992-10-01)
The cell wall-associated proteinase from Lactococcus lactis subsp. cremoris H2 (isolate number 4409) was released from the cells by treatment with lysozyme, even in the presence of 50 mM calcium chloride. Cell lysis during lysozyme treatment was minimal. The proteinase
R.W. Franck
Bioorganic Chemistry, 20, 77-77 (1992)
Infrared absorbance spectroscopy of aqueous proteins: Comparison of transmission and ATR data collection and analysis for secondary structure fitting
Corujo MP, et al.
Chirality, 30(8), 957-965 (2018)
L. Stevens
Comp. Biochem. Physiol., B: Comp. Biochem., 100, 1-1 (1991)
Structures and interactions among lysozyme proteins below the isoelectric point in presence of divalent ions
Pandit S, et al.
Chemical Physics Letters, 711, 8-14 (2018)

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service