Skip to Content
Merck
All Photos(1)

Key Documents

SML1525

Sigma-Aldrich

(-)-JQ1

≥95% (HPLC)

Synonym(s):

(R)-(-)-tert-Butyl 2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetate, 6H-Thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetic acid, 4-(4-chlorophenyl)-2,3,9-trimethyl-, 1,1-dimethylethyl ester, (6R)-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$259.00
25 MG
$939.00

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$259.00
25 MG
$939.00

About This Item

Empirical Formula (Hill Notation):
C23H25ClN4O2S
CAS Number:
Molecular Weight:
456.99
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC(C)(C)C)C[C@@H]1C2=NN=C(C)N2C(SC(C)=C3C)=C3C(C4=CC=C(Cl)C=C4)=N1

InChI

1S/C23H25ClN4O2S/c1-12-13(2)31-22-19(12)20(15-7-9-16(24)10-8-15)25-17(11-18(29)30-23(4,5)6)21-27-26-14(3)28(21)22/h7-10,17H,11H2,1-6H3/t17-/m1/s1

InChI key

DNVXATUJJDPFDM-QGZVFWFLSA-N

General description

JQ1 is a member of the triazolo-diazepine compound family which functions as a pan-BET (bromodomain and extra-terminal motif) family inhibitor.[1] Its core structure is composed of thienodiazepine.[2]

Biochem/physiol Actions

(-)-JQ1 is an inactive enantiomer of JQ1 that serves as a negative control.
(-)-JQ1 is the negative control probe for (+)-JQ1, the active enantiomer of (+/-)-JQ1, which is a potent and selective BET bromodomain inhibitor. (-)-JQ1 shows no significant interaction with BRD1-4 or other bromodomains.

For characterization details of the active probe, (+)-JQ1, please visit the JQ1 probe summary on the Structural Genomics Consortium (SGC) website.

(+)-JQ1, the active enantiomer, is available from Sigma. To learn more about and purchase (+)-JQ1, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

(-) JQ-1 is a negative control for (+) JQ1, an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting chromatin readers.
James LI and Frye SV
Clinical Pharmacology and Therapeutics, 93(4), 312-314 (2013)
Epigenetic chemical probes.
Muller S and Brown PJ
Clinical Pharmacology and Therapeutics, 92(6), 689-693 (2012)
Jiwoon Lee et al.
Glia, 71(12), 2866-2883 (2023-08-16)
The zebrafish retina possesses tremendous regenerative potential. Müller glia underlie retinal regeneration through their ability to reprogram and generate multipotent neuronal progenitors that re-differentiate into lost neurons. Many factors required for Müller glia reprogramming and proliferation have been identified; however
Tatsuro Yamamoto et al.
British journal of cancer, 123(4), 580-590 (2020-06-06)
Oral squamous cell carcinoma (OSCC) has increased morbidity, and its high metastatic potential affects patient survival. Bromodomain containing 4 (BRD4) is a chromatin protein that associates with acetylated histone lysines and facilitates transcription. BRD4 has been implicated in cell proliferation
Hilmar Strickfaden et al.
Cell, 183(7), 1772-1784 (2020-12-17)
The association of nuclear DNA with histones to form chromatin is essential for temporal and spatial control of eukaryotic genomes. In this study, we examined the physical state of condensed chromatin in vitro and in vivo. Our in vitro studies demonstrate that self-association

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service