Skip to Content
Merck
All Photos(1)

Key Documents

SML2623

Sigma-Aldrich

(+)-JQ-1 carboxylic acid

≥95% (HPLC)

Synonym(s):

(6S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetic acid, (S)-[4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-yl]acetic acid, 2-[(6S,Z)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl]acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$170.00
25 MG
$674.00

$170.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$170.00
25 MG
$674.00

About This Item

Empirical Formula (Hill Notation):
C19H17ClN4O2S
CAS Number:
Molecular Weight:
400.88
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

$170.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1C)C)C(=N[C@H](c4[n]2c(nn4)C)CC(=O)O)c3ccc(cc3)Cl

InChI

1S/C19H17ClN4O2S/c1-9-10(2)27-19-16(9)17(12-4-6-13(20)7-5-12)21-14(8-15(25)26)18-23-22-11(3)24(18)19/h4-7,14H,8H2,1-3H3,(H,25,26)/t14-/m0/s1

InChI key

LJOSBOOJFIRCSO-AWEZNQCLSA-N

Biochem/physiol Actions

(+)-JQ-1 carboxylic acid is a potent BET (bromodomain and extra terminal domain) inhibitor. (+)-JQ-1 carboxylic acid could serve as precursor to synthesize PROTACs and other conjugates.
potent BET inhibitor; precursor to synthesize PROTACs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morgan S Gadd et al.
Nature chemical biology, 13(5), 514-521 (2017-03-14)
Inducing macromolecular interactions with small molecules to activate cellular signaling is a challenging goal. PROTACs (proteolysis-targeting chimeras) are bifunctional molecules that recruit a target protein in proximity to an E3 ubiquitin ligase to trigger protein degradation. Structural elucidation of the
John Hines et al.
Cancer research, 79(1), 251-262 (2018-11-06)
Although the number of proteins effectively targeted for posttranslational degradation by PROTAC has grown steadily, the number of E3 ligases successfully exploited to accomplish this has been limited to the few for which small-molecule ligands have been discovered. Although the
Georg E Winter et al.
Science (New York, N.Y.), 348(6241), 1376-1381 (2015-05-23)
The development of effective pharmacological inhibitors of multidomain scaffold proteins, notably transcription factors, is a particularly challenging problem. In part, this is because many small-molecule antagonists disrupt the activity of only one domain in the target protein. We devised a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service