Skip to Content
Merck
All Photos(1)

Key Documents

C6374

Sigma-Aldrich

Caerulomycin A

from Streptomyces caeruleus, ≥98% (HPLC)

Synonym(s):

Carulomycin A, Cerulomycin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$564.00

$564.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$564.00

About This Item

Empirical Formula (Hill Notation):
C12H11N3O2
CAS Number:
Molecular Weight:
229.23
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$564.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

biological source

Streptomyces caeruleus

Quality Level

Assay

≥98% (HPLC)

form

powder

antibiotic activity spectrum

fungi

storage temp.

−20°C

SMILES string

COc1cc(\C=N\O)nc(c1)-c2ccccn2

InChI

1S/C12H11N3O2/c1-17-10-6-9(8-14-16)15-12(7-10)11-4-2-3-5-13-11/h2-8,16H,1H3/b14-8+

InChI key

JCTRJRHLGOKMCF-RIYZIHGNSA-N

Biochem/physiol Actions

Caerulomycins produced by Streptomyces caeruleus are bipyridinic molecules endowed with antibiotic properties. Caerulomycin A was found to have a strong anti-fungal and anti-amoebic activity, and a mild antibacterial activity.

Preparation Note

Caerulomycin A stock solution may be prepared in DMSO or methanol at 1 mg/mL. The stock solution can be further diluted in water to 0.1 mg/mL. Higher concentrations my requrie gentle heating to 40 °C. Dilutions from the stock solution should be freshly prepared.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D K Chatterjee et al.
Zeitschrift fur Parasitenkunde (Berlin, Germany), 70(5), 569-573 (1984-01-01)
The anti-amoebic action of the bipyridyl antibiotic caerulomycin was assessed in vitro and in vivo using various strains of Entamoeba histolytica from polyxenic, axenic and monoxenic cultures. Minimum inhibition concentrations of caerulomycin (metronidazole) were 7.5 (5), 15.6(1.95) and 60 (2.5)
Tarek Sammakia et al.
Organic letters, 4(14), 2385-2388 (2002-07-06)
[reaction: see text] The total synthesis of caerulomycin C is described. Key steps in this synthesis utilize 1,2-, 1,3-, and 1,4-halogen dance reactions for the functionalization of the pyridine ring.
Qinheng Lin et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 51(8), 1032-1041 (2011-11-22)
In order to enable the caerulomyicn biosynthetic study by in vivo gene disruptions, it is crucial to develop a genetic modification system for the producer Actinoalloteichus sp. WH1-2216-6. The spore germination timing and the concentration of MgSO4 in the medium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service