Skip to Content
Merck
All Photos(1)

Key Documents

SML1483

Sigma-Aldrich

Ogerin negative control

≥98% (HPLC)

Synonym(s):

3-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]-benzenemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00
25 MG
$465.00

$115.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$115.00
25 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C17H17N5O
CAS Number:
Molecular Weight:
307.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(C2=CC(CO)=CC=C2)=NC(NCC3=CC=CC=C3)=N1

InChI

1S/C17H17N5O/c18-16-20-15(14-8-4-7-13(9-14)11-23)21-17(22-16)19-10-12-5-2-1-3-6-12/h1-9,23H,10-11H2,(H3,18,19,20,21,22)

InChI key

RVKAHYFWSKSSQQ-UHFFFAOYSA-N

Biochem/physiol Actions

Ogerin negative control is a structurally similar analog of ogerin (catalog no. SML1482). Ogerin is a selective positive allosteric modulator of an orphan GPCR, GPR68, also known as Ovarian cancer G-protein coupled receptor 1 (OGR1). GPR68 is one of the proton or pH-sensing GPCRs that sense extracellular H(+). Ogerin potently potentiated proton-mediated GPR68-Gs signaling. The meta-analog is inactive and serves as a negative control.

Recommended products

The positive allosteric modulator, ogerin, is available from Sigma. To learn more about and purchase ogerin, click here.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service