Skip to Content
Merck
All Photos(3)

Key Documents

N2255

Sigma-Aldrich

S-(4-Nitrobenzyl)-6-thioinosine

≥98%, solid

Synonym(s):

6-[(4-Nitrobenzyl)thio]-9-β-D-ribofuranosylpurine, NBMPR, NBTI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N5O6S
CAS Number:
Molecular Weight:
419.41
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
form:
solid
Assay:
≥98%

Quality Level

Assay

≥98%

form

solid

color

white

mp

187-190 °C (lit.)

solubility

0.1 M HCl: slightly soluble
0.1 M NaOH: slightly soluble
DMSO: soluble
H2O: insoluble

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(SCc4ccc(cc4)[N+]([O-])=O)ncnc23

InChI

1S/C17H17N5O6S/c23-5-11-13(24)14(25)17(28-11)21-8-20-12-15(21)18-7-19-16(12)29-6-9-1-3-10(4-2-9)22(26)27/h1-4,7-8,11,13-14,17,23-25H,5-6H2/t11-,13-,14-,17-/m1/s1

InChI key

DYCJFJRCWPVDHY-LSCFUAHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-(4-Nitrobenzyl)-6-thioinosine (NBTI) belongs to the family of S6-substituted 6-thiopurine nucleosides, which regulate nucleoside transport mechanisms in animals. It acts as a ligand of adenosine transporter. Binding sites for NBTI is located on brain capillaries. It functions as a covalent photoaffinity probe for nucleoside transport.

Biochem/physiol Actions

Inhibitor of equilibrative nucleoside transporters (ENTs), particularly adenosine transporters, in central nervous system and vascular smooth muscle.
Potent adenosine uptake inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulatory Function of Adenosine, 268(1), 14-18 (2012)
Sophie M Stief et al.
Leukemia, 34(1), 50-62 (2019-06-16)
Acute myeloid leukemia (AML) is an aggressive hematologic neoplasm resulting from the malignant transformation of myeloid progenitors. Despite intensive chemotherapy leading to initial treatment responses, relapse caused by intrinsic or acquired drug resistance represents a major challenge. Here, we report
Blood-brain barrier transport and brain metabolism of adenosine and adenosine analogs.
Pardridge WM, et al.
Journal of Pharmacology and Experimental Therapeutics, 268(1), 14-18 (1994)
Morris J Robins et al.
Journal of medicinal chemistry, 53(16), 6040-6053 (2010-08-20)
5'-S-(2-aminoethyl)-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAENTA), 5'-S-(2-acetamidoethyl)-6-N-[(4-substituted)benzyl]-5'-thioadenosine analogues, 5'-S-[2-(6-aminohexanamido)]ethyl-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAHENTA), and related compounds were synthesized by S(N)Ar displacement of fluoride from 6-fluoropurine intermediates with 4-(substituted)benzylamines. Conjugation of the pendant amino groups of SAENTA and SAHENTA with fluorescein-5-yl isothiocyanate (FITC) gave fluorescent probes that bound
Methods Used in Adenosine Research, 268(1), 14-18 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service