Skip to Content
Merck
All Photos(1)

Key Documents

SML1448

Sigma-Aldrich

Cycloastragenol

≥98% (HPLC)

Synonym(s):

Astramembrangenin, Cyclosieversigenin, Cyclosiversigenin, GRN510, TAT2, (3β,6α,16β,20R,24S)-20,24-Epoxy-9,19-cyclolanostane-3,6,16,25-tetrol, Cyclogalegigenin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$464.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$144.00
25 MG
$464.00

About This Item

Empirical Formula (Hill Notation):
C30H50O5
CAS Number:
Molecular Weight:
490.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$144.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +45 to +60°, c = 1 in methanol

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@]1(O[C@H](C(O)(C)C)CC1)[C@@]2([H])[C@@H](O)C[C@@]3(C)[C@]4([H])C[C@H](O)[C@@]5([H])C(C)(C)[C@@H](O)CC[C@]5(C6)[C@]46CC[C@@]32C

InChI

1S/C30H50O5/c1-24(2)20(33)8-11-30-16-29(30)13-12-26(5)23(28(7)10-9-21(35-28)25(3,4)34)18(32)15-27(26,6)19(29)14-17(31)22(24)30/h17-23,31-34H,8-16H2,1-7H3/t17-,18-,19-,20-,21-,22-,23-,26+,27-,28+,29-,30+/m0/s1

InChI key

WENNXORDXYGDTP-UOUCMYEWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cycloastragenol is isolated from Astragalus species.

Biochem/physiol Actions

Cycloastragenol is an aglycone of astragaloside IV isolated from Astragalus species that exhibits anti-inflammatory properties and promotes and wound gap closure. Cycloastragenol enhances the antiviral response in human CD8+ T-lymphocytes. Cycloastragenol induces telomerase activity and induces CREB (cAMP response element binding) activation in human neonatal keratinocytes, PC12 cells, and primary neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanny C F Ip et al.
Neuro-Signals, 22(1), 52-63 (2014-08-07)
Cycloastragenol (CAG) is an aglycone of astragaloside IV. It was first identified when screening Astragalus membranaceus extracts for active ingredients with antiaging properties. The present study demonstrates that CAG stimulates telomerase activity and cell proliferation in human neonatal keratinocytes. In
Claude Jourdan Le Saux et al.
PloS one, 8(3), e58423-e58423 (2013-03-22)
The emergence of diseases associated with telomere dysfunction, including AIDS, aplastic anemia and pulmonary fibrosis, has bolstered interest in telomerase activators. We report identification of a new small molecule activator, GRN510, with activity ex vivo and in vivo. Using a
Steven Russell Fauce et al.
Journal of immunology (Baltimore, Md. : 1950), 181(10), 7400-7406 (2008-11-05)
Telomerase reverse transcribes telomere DNA onto the ends of linear chromosomes and retards cellular aging. In contrast to most normal somatic cells, which show little or no telomerase activity, immune cells up-regulate telomerase in concert with activation. Nevertheless, during aging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service