Skip to Content
Merck
All Photos(2)

Key Documents

SMB00372

Sigma-Aldrich

Cycloastragenol

≥98% (HPLC)

Synonym(s):

(3β,6α,16β,20R,24S)-20,24-Epoxy-9,19-cyclolanostane-3,6,16,25-tetrol, Astramembrangenin, Cyclogalegigenin, Cyclosieversigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O5
CAS Number:
Molecular Weight:
490.72
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@]1(O[C@H](C(O)(C)C)CC1)[C@@]2([H])[C@@H](O)C[C@@]3(C)[C@]4([H])C[C@H](O)[C@@]5([H])C(C)(C)[C@@H](O)CC[C@]5(C6)[C@]46CC[C@@]32C

InChI

1S/C30H50O5/c1-24(2)20(33)8-11-30-16-29(30)13-12-26(5)23(28(7)10-9-21(35-28)25(3,4)34)18(32)15-27(26,6)19(29)14-17(31)22(24)30/h17-23,31-34H,8-16H2,1-7H3/t17-,18-,19-,20-,21-,22-,23-,26+,27-,28+,29-,30+/m0/s1

InChI key

WENNXORDXYGDTP-UOUCMYEWSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SML1448PHL82509PHL80357
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98.0% (HPLC)

assay

≥98.0% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

-

application(s)

food and beverages

application(s)

-

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

General description

Cycloastragenol is a sapogenin and an aglycone found in or derived from Astragalus membranaceus.[1] It is a natural triterpenoid.[1]

Biochem/physiol Actions

Cycloastragenol is used as a nutraceutical (e.g. TAT2) and seems to moderately increase the telomerase activity and proliferative capacity of both CD4 and CD8 T-cells. Cycloastragenol possesses antiviral, antimicrobial, anti-inflammatory, anti-hepatotoxic, anti-leukemia, antitumor and antinociceptive activity. It also acts as an immunomodulator.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro intestinal absorption and first-pass intestinal and hepatic metabolism of cycloastragenol, a potent small molecule telomerase activator
Zhu J, et al.
Drug Metabolism and Pharmacokinetics, 25(5), 477-486 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service